Merck
  • Investigation of the structure-selectivity relationships and van't Hoff analysis of chromatographic stereoisomer separations of unusual isoxazoline-fused 2-aminocyclopentanecarboxylic acids on Cinchona alkaloid-based chiral stationary phases.
Investigation of the structure-selectivity relationships and van't Hoff analysis of chromatographic stereoisomer separations of unusual isoxazoline-fused 2-aminocyclopentanecarboxylic acids on Cinchona alkaloid-based chiral stationary phases.
Journal of chromatography. A (2015-02-11)
István Ilisz, Zsanett Gecse, Gyula Lajkó, Melinda Nonn, Ferenc Fülöp, Wolfgang Lindner, Antal Péter
ABSTRACT
The enantiomers of four unusual, rather rigid isoxazoline-fused 2-aminocyclopentanecarboxylic acids were directly separated on a quinine- or a quinidine-based zwitterionic ion-exchanger as chiral selector. The effects of the mobile phase composition, the structures of the analytes and temperature on the separations were investigated. Experiments were performed at constant mobile phase composition in the temperature range 10-50°C to study the effects of temperature, and thermodynamic parameters were calculated from plots of ln α versus 1/T. Some mechanistic aspects of the chiral recognition process are discussed with respect to the structures of the analytes. It was found that the enantiomer separations were in most cases predominantly enthalpy-driven, but entropically-driven separations were also observed. The sequence of elution of the enantiomers was determined in all cases.
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
Triethylamine, ≥99.5%
Supelco
Acetic acid, analytical standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Butylamine, ≥99%
Sigma-Aldrich
N,N-Diethylaniline, ≥99%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Tripropylamine, ≥98%
Sigma-Aldrich
Butylamine, 99.5%
Sigma-Aldrich
Propylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Propylamine, 98%
Sigma-Aldrich
Propylamine, purum, ≥99.0% (GC)
Sigma-Aldrich
Triethylamine, purum, ≥99% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Ammonia solution, 2.0 M in methanol