Merck
  • New pressure-assisted sweeping on-line preconcentration for highly polar environmentally relevant nitrosamines: Part 2. Cationic and anionic surfactants with zero-flow capillaries.
New pressure-assisted sweeping on-line preconcentration for highly polar environmentally relevant nitrosamines: Part 2. Cationic and anionic surfactants with zero-flow capillaries.
Electrophoresis (2015-02-03)
Ali Modir-Rousta, Christina S Bottaro
ABSTRACT
We recently introduced a pressure-assisted sweeping-reversed migration-EKC (RM-EKC) method for preconcentration of neutral polar N-nitrosamines with low affinity for the micellar phase. The type of surfactant and phase ratio are dominant factors in dictating the magnitude of interactions between analyte and micellar phase, thus four surfactants (anionic and cationic) with a range of functionalities (SDS, ammonium perfluorooctanoate (APFO), bile salts, and cetyltrimethylammonium chloride (CTAC)) were evaluated for sweeping-RM-EKC of highly polar N-nitrosamines. All gave acceptable results for sweeping-RM-EKC when used in high concentrations (≥200 mM) with low EOF. While no single surfactant was superior by all measures, all but the bile salts had useful performance characteristics. APFO showed the narrowest peak widths and highest number of theoretical plates, though two species co-migrated at all concentrations (25-300 mM); SDS and the cationic surfactant CTAC also showed good separation characteristics and could resolve all peaks, but CTAC had wider separation window. Various types of capillaries coated for EOF control were compared for use with anionic and cationic surfactants. A commercial zero-EOF capillary coated with a polymer bearing sulfonic acid functional groups showed superior EOF suppression and reproducibility of migration time with all surfactants.
Product Description
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
USP
Cetrimonium bromide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Pentadecafluorooctanoic acid ammonium salt, ≥98.0% (NT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Dodecanophenone, 98%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%