Merck
  • Controlled release studies of antimalarial 1, 3, 5-trisubstituted-2-pyrazolines from biocompatible chitosan-heparin Layer-by-Layer (LbL) self assembled thin films.
Controlled release studies of antimalarial 1, 3, 5-trisubstituted-2-pyrazolines from biocompatible chitosan-heparin Layer-by-Layer (LbL) self assembled thin films.
Colloids and surfaces. B, Biointerfaces (2014-12-09)
Uma M Bhalerao, Aditya Kapil Valiveti, Jyotiranjan Acharya, Anand K Halve, Mahabir Parshad Kaushik
ABSTRACT
Herein we report the in-vitro controlled release properties of 1, 3, 5-trisubstituted-2-pyrazolines through Layer-by-Layer (LbL) self assembled thin films fabricated from chitosan and heparin sodium salt as biocompatible polyelectrolytes. This study was carried out as a preliminary step towards the applicability of LbL technique in prophylactic drug delivery of antimalarial drugs. The growth of LbL self assembly was monitored by UV-Visible spectrophotometry and Quartz Crystal Microbalance (QCM). The loading as well as in-vitro release studies (in phosphate buffer saline at pH 7.4) were carried out using UV-Visible spectroscopy. Three compounds having good antimalarial activity were tested and the release rate was found inversely proportional to the hydrophobicity of the drug. Pzln-4 has shown best release among all the three compounds (up to 780 min) followed by Pzln-5 and Pzln-8. The release trend was that of a fast release up to first 2 h followed by a steady release. Kinetic fitting of the data confirmed the process of drug release followed a pseudo second order kinetics (R(2)≥0.99). A large value of rate constant (k) revealed a faster release. Pzln-4 has shown smallest value of k corresponding to slowest release among all the three compounds.
Product Description
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
2-Propen-1-amine hydrochloride, AldrichCPR
Sigma-Aldrich
3-Mercaptopropionic acid, ≥99.0% (HPLC)
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
3-Mercaptopropionic acid, ≥99%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Zirconyl chloride octahydrate, reagent grade, 98%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Nicotinic hydrazide, 97%
Sigma-Aldrich
Allylamine hydrochloride, 98%