Merck
  • Lipophilicity of amine neurotransmitter precursors, metabolites and related drugs estimated on various TLC plates.
Lipophilicity of amine neurotransmitter precursors, metabolites and related drugs estimated on various TLC plates.
Journal of chromatographic science (2013-10-29)
Simona Codruţa Cobzac, Dorina Casoni, Costel Sârbu
ABSTRACT
The retention behavior for a series of amine neurotransmitters, their precursors, metabolites and structurally related drugs has been investigated in reversed-phase thin-layer chromatography using RP-18, RP-8, RP-2, CN and Diol stationary phases and mixtures of phosphate buffer and methanol as mobile phase. According to the computed lipophilicity of the neutral form of the investigated compounds, the most lipophilic compound is dobutamine (log P(N) = 3.78), while the less lipophilic is norepinephrine (log P(N) = -0.14). The experimental results also show dobutamine as the most lipophilic compound in the case of RP-18 and CN stationary phases (RM0(RP-18) = 1.58 and RM(CN) = 1.21), while RP-8 indicates norepinephrine as the less lipophilic one (RM0(RP8) = -0.70). Both the theoretical computation and the experimental data revealed that only one ionic form of the compounds prevails in the used chromatographic conditions. In addition, the evaluation of the experimental results showed that a similar chromatographic behavior could be assumed in the case of RP-18, RP-8 and CN stationary phases. Moreover, the mRM (mean of the RM values) and PC1/RM (scores of the first principal component) experimental lipophilicity indices showed a high correlation with the computed lipophilicity indices.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Salbutamol sulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
Dopamine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Phenylephrine Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Albuterol sulfate, Pharmaceutical Secondary Standard; Certified Reference Material
Dopamine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
DL-4-Hydroxy-3-methoxymandelic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Methoxamine hydrochloride
Sigma-Aldrich
Dopamine hydrochloride
Sigma-Aldrich
Metaproterenol hemisulfate salt
Phenylephrine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
3,4-Dihydroxyphenylacetic acid, 98%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Homovanillic acid, Fluorimetric reagent
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
USP
Phenylephrine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%