Skip to Content
Merck
All Photos(1)

Key Documents

D2960000

Dopamine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-(3,4-Dihydroxyphenyl)ethylamine hydrochloride, 3,4-Dihydroxyphenethylamine hydrochloride, 3-Hydroxytyramine hydrochloride, 4-(2-Aminoethyl)-1,2-benzenediol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CH2CH2NH2·HCl
CAS Number:
Molecular Weight:
189.64
Beilstein:
3656720
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

dopamine

manufacturer/tradename

EDQM

mp

248-250 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

SMILES string

Cl[H].NCCc1ccc(O)c(O)c1

InChI

1S/C8H11NO2.ClH/c9-4-3-6-1-2-7(10)8(11)5-6;/h1-2,5,10-11H,3-4,9H2;1H

InChI key

CTENFNNZBMHDDG-UHFFFAOYSA-N

Gene Information

human ... ADRB1(153)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dopamine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adriana Basnakova et al.
Neurobiology of stress, 15, 100403-100403 (2021-10-12)
The response of an animal to a sensory stimulus depends on the nature of the stimulus and on expectations, which are mediated by spontaneous activity. Here, we ask how circadian variation in the expectation of danger, and thus the response
Frank J Meye et al.
Trends in pharmacological sciences, 35(1), 31-40 (2013-12-18)
Overconsumption of high caloric food plays an important role in the etiology of obesity. Several factors drive such hedonic feeding. High caloric food is often palatable. In addition, when an individual is sated, stress and food-related cues can serve as
Hanneke E M den Ouden et al.
Neuron, 80(4), 1090-1100 (2013-11-26)
Serotonin and dopamine are speculated to subserve motivationally opponent functions, but this hypothesis has not been directly tested. We studied the role of these neurotransmitters in probabilistic reversal learning in nearly 700 individuals as a function of two polymorphisms in
Sebastian C Holst et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(2), 566-573 (2014-01-10)
While dopamine affects fundamental brain processes such as movement control, emotional responses, addiction, and pain, the roles for this neurotransmitter in regulating wakefulness and sleep are incompletely understood. Genetically modified animal models with reduced dopamine clearance exhibit hypersensitivity to caffeine
Horng H Chen et al.
JAMA, 310(23), 2533-2543 (2013-11-20)
Small studies suggest that low-dose dopamine or low-dose nesiritide may enhance decongestion and preserve renal function in patients with acute heart failure and renal dysfunction; however, neither strategy has been rigorously tested. To test the 2 independent hypotheses that, compared

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service