Merck
  • Potential amelioration of upregulated renal HIF-1alpha-endothelin-1 system by landiolol hydrochloride in a rat model of endotoxemia.
Potential amelioration of upregulated renal HIF-1alpha-endothelin-1 system by landiolol hydrochloride in a rat model of endotoxemia.
Life sciences (2014-05-23)
Yoshiyasu Ogura, Subrina Jesmin, Naoto Yamaguchi, Masami Oki, Nobutake Shimojo, Md Majedul Islam, Tanzila Khatun, Junko Kamiyama, Hideaki Sakuramoto, Keiichi Hagiya, Satoru Kawano, Taro Mizutani
ABSTRACT
Endothelin (ET)-1 is the best known potent vasoconstrictor and has been implicated in pathogenesis of sepsis-associated acute kidney injury (AKI) in human or lipopolysaccharide (LPS)-induced AKI in animal models. We have previously shown that ET-1 is highly up-regulated in renal tissues and in plasma after LPS administration. Here, we investigated whether landiolol hydrochloride, an ultra-short-acting beta-blocker, can play an important role in ameliorating levels of LPS-induced up-regulation of renal HIF-1α-ET-1 system and inflammatory cytokines in a rat model of endotoxemia. Male Wistar rats at 8 weeks of age were either administered with: a) lipopolysaccharide (LPS) only for three hours (3 h) or b) LPS, followed by continuous administration of landiolol for 3 h; c) third group was only treated with vehicle. At 3 h after LPS administration there was: a) minimal injury in kidney tissues; b) circulatory levels of creatinine, blood urea nitrogen and NGAL increased and c) expression of inflammatory cytokines, such as TNF-α, IL-6 and iNOS increased at the level of both circulatory and renal tissues. In addition, LPS significantly induced renal expression of ET-1 and HIF-1α compared to control. Finally, treatment of LPS-administered rats with landiolol for 3 h normalized elevated serum markers of renal injury and up-regulated levels of renal HIF-1α-ET-1 system with normalization of TNF-α. Taken together, these data led us to conclude that landiolol ameliorates the up-regulation of HIF-1α-ET-1 system in minimally morphologically-injured kidney and normalizes biomarkers of renal injury in early hours of endotoxemia of a rat model.
Product Description
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Creatine, anhydrous
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
USP
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Urea, analytical standard
Urea, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Landiolol hydrochloride, ≥98% (HPLC)