Merck
  • Determination of low levels of benzodiazepines and their metabolites in urine by hollow-fiber liquid-phase microextraction (LPME) and gas chromatography-mass spectrometry (GC-MS).
Determination of low levels of benzodiazepines and their metabolites in urine by hollow-fiber liquid-phase microextraction (LPME) and gas chromatography-mass spectrometry (GC-MS).
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-12-03)
André Valle de Bairros, Rafael Menck de Almeida, Lorena Pantaleão, Thiago Barcellos, Sidnei Moura e Silva, Mauricio Yonamine
ABSTRACT
In this study, it is shown a method for the determination of benzodiazepines and their main metabolites in urine samples by hollow-fiber liquid-phase microextraction (LPME) in the three-phase mode. Initially, the hydrolysis step was performed using 100 μL of sodium acetate 2.0 mol/L buffer solution (pH 4.5), 25 μL of β-glucuronidase enzyme and incubation for 90 min at 55 °C. In parallel with hydrolysis, the LPME fiber (9 cm) was prepared. Its pores were filled with a mixture of dihexyl ether: 1-nonanol (9:1). Afterwards, a solution of 3.0 mol/L of HCl was introduced into the lumen of the fiber (acceptor phase). After hydrolysis, the fiber was submersed in the alkalinized urine (pH 10) containing 10% NaCl. Samples were then submitted to orbital shaking (2400 rpm) for 90 min. The acceptor phase was later withdrawn from the fiber, dried and the residue derivatized with trifluoroacetic anhydride (TFAA) for 10 min at 60 °C with further addition of N-methyl-N-tert-butyldimethylsilyltrifluoroacetamide containing 1% tert-butyldimethylchlorosilane (MTBSTFA) for 45 min at 90 °C followed by determination by gas chromatography-mass spectrometry (GC-MS). The calibration curves obtained showed linearity over the specified range, with a similar sensitivity to traditional techniques and a higher detection capability compared to most of the miniaturized methods described in the literature. The method has been developed and successfully validated and applied to urine samples from real cases of benzodiazepines intake.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Supelco
Clonazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Nitrazepam
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Supelco
Nordiazepam solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
tert-Butylchlorodimethylsilane solution, 50 wt. % in toluene
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Supelco
Undecane, analytical standard
Sigma-Aldrich
1-Octanol, ≥98%, FCC, FG
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Supelco
Trifluoroacetic anhydride, for GC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Undecane, ≥99%
Sigma-Aldrich
Trifluoroacetic anhydride, ReagentPlus®, ≥99%