Merck
  • Direct detection of free fatty acids in edible oils using supercritical fluid chromatography coupled with mass spectrometry.
Direct detection of free fatty acids in edible oils using supercritical fluid chromatography coupled with mass spectrometry.
Food chemistry (2014-10-13)
Shuping Qu, Zhenxia Du, Yun Zhang
ABSTRACT
Determination of free fatty acids (FFAs) in food products is of enormous interest mainly because they are related to the quality and authenticity of the oils. In this study, supercritical fluid chromatography (SFC), followed by an electrospray ionisation triple-quadrupole mass spectrometry (ESI-MS), is shown to provide a novel method for the separation and detection of FFAs in edible oils without any pretreatment. Eight FFAs were separated on a HSS C18 SB column with gradient elution within 3 min. Effects of different columns, modifiers and column temperature changes were evaluated. The results indicated the feasibility of this method for the high-throughput determination of individual FFAs with satisfactory correlation coefficients (R(2)>0.994) and good reproducibility of RSD<13.5% (intraday) and <15.0% (interday). By combined with principal component analysis (PCA), different types of edible oil were successfully distinguished into several categories, showing a potential application for the determination of oil quality or authenticity.
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Supelco
Stearic acid, analytical standard
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
Oleic acid, natural, FCC
Supelco
Oleic acid, Selectophore, ≥99%
Stearic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Hexane, analytical standard
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%