Merck
  • Modeling nociception in zebrafish: a way forward for unbiased analgesic discovery.
Modeling nociception in zebrafish: a way forward for unbiased analgesic discovery.
PloS one (2015-01-15)
Andrew Curtright, Micaela Rosser, Shamii Goh, Bailey Keown, Erinn Wagner, Jasmine Sharifi, David W Raible, Ajay Dhaka
ABSTRACT
Acute and chronic pain conditions are often debilitating, inflicting severe physiological, emotional and economic costs and affect a large percentage of the global population. However, the development of therapeutic analgesic agents based primarily on targeted drug development has been largely ineffective. An alternative approach to analgesic development would be to develop low cost, high throughput, untargeted animal based behavioral screens that model complex nociceptive behaviors in which to screen for analgesic compounds. Here we describe the development of a behavioral based assay in zebrafish larvae that is effective in identifying small molecule compounds with analgesic properties. In a place aversion assay, which likely utilizes supraspinal neuronal circuitry, individually arrayed zebrafish larvae show temperature-dependent aversion to increasing and decreasing temperatures deviating from rearing temperature. Modeling thermal hyperalgesia, the addition of the noxious inflammatory compound and TRPA1 agonist allyl isothiocyanate sensitized heat aversion and reversed cool aversion leading larvae to avoid rearing temperature in favor of otherwise acutely aversive cooler temperatures. We show that small molecules with known analgesic properties are able to inhibit acute and/or sensitized temperature aversion.
Product Description
Supelco
Ibuprofen
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
Clonidine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard
Pancuronium bromide, European Pharmacopoeia (EP) Reference Standard
Supelco
Promethazine hydrochloride, vial of 25 mg, certified reference material, Cerilliant®
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Supelco
Benzoylecgonine-d3 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Pancuronium bromide
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Promethazine hydrochloride
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Clonidine hydrochloride, solid
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Amitriptyline hydrochloride, ≥98% (TLC), powder
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Ethyl 3-aminobenzoate methanesulfonate, 98%
Promethazine hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Ibuprofen, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
USP
Amitriptyline hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Ethyl 3-aminobenzoate methanesulfonate salt, analytical standard