Skip to Content
Merck

Skip To

I110

Ibuprofen

Synonym(s):

α-Methyl-4-(isobutyl)phenylacetic acid, (±)-2-(4-Isobutylphenyl)propanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C13H18O2
CAS Number:
Molecular Weight:
206.28
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
239-784-6
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

Quality Segment

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

color

white to off-white

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CC(C)Cc1ccc(cc1)C(C)C(O)=O

InChI

1S/C13H18O2/c1-9(2)8-11-4-6-12(7-5-11)10(3)13(14)15/h4-7,9-10H,8H2,1-3H3,(H,14,15)

InChI key

HEFNNWSXXWATRW-UHFFFAOYSA-N

Gene Information

General description

Ibuprofen is a non-steroidal and anti-inflammatory drug, which finds a variety of therapeutic applications in relieving pain and inflammatory disorders.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Ibuprofen has been used as a test compound in studying its removal from aqueous solutions via adsorption on titanium dioxide based multi-walled carbon nanotube membranes. It is also used as an external reference standard in order to validate the purity of the macro-cyclic peptides, an emerging therapeutic drug modality, using nuclear magnetic resonance technique (NMR).

Biochem/physiol Actions

Cyclooxygenase (COX) inhibitor that has greater activity against COX-1 than against COX-2.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR10041335508I0020000
description

-

description

Pharmaceutical Secondary Standard; Certified Reference Material

description

United States Pharmacopeia (USP) Reference Standard

description

European Pharmacopoeia (EP) Reference Standard

format

neat

format

neat

format

neat

format

neat

assay

≥98% (HPLC)

assay

-

assay

-

assay

-

technique(s)

gas chromatography (GC): suitable, HPLC: suitable

technique(s)

gas chromatography (GC): suitable, HPLC: suitable

technique(s)

-

technique(s)

-

Quality Level

100

Quality Level

300

Quality Level

-

Quality Level

-

color

white to off-white

color

-

color

-

color

-

application(s)

forensics and toxicology
pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)


Still not finding the right product?

Explore all of our products under Ibuprofen


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters