Merck
  • Method validation and application of a liquid chromatography-tandem mass spectrometry method for drugs of abuse testing in exhaled breath.
Method validation and application of a liquid chromatography-tandem mass spectrometry method for drugs of abuse testing in exhaled breath.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-02-18)
Niclas Stephanson, Sören Sandqvist, Marjan Shafaati Lambert, Olof Beck
ABSTRACT
A mass spectrometric method for drugs of abuse testing in exhaled breath employing a sampling device collecting aerosol particles was developed and applied in routine use. Analytes covered were amphetamine, methamphetamine, 6-acetylmorphine, morphine, cocaine, benzoylecgonine, diazepam, oxazepam and tetrahydrocannabinol. The method involved eluting drugs from the collection filter with methanol, quantification using deuterated analogs as internal standards, reversed phase chromatography with gradient elution, positive electrospray ionization and monitoring of two product ions per analyte in selected reaction monitoring mode. The measuring range was 6.0-1000pg/filter. The intra- and inter-assay imprecision expressed as the coefficient of variation was less than 7%. Influence from matrix was noted for most compounds but was compensated for the use of co-eluting internal standards. The LLOQ was 6.0pg/filter with intra-assay CV <5% and accuracy within 99-102% for all analytes. No chromatographic interference was observed in 20 negative control samples. The LC-MS/MS method was successfully applied for measuring drugs in unknown samples collected for the purpose of drug testing. Among the 1096 analyzed samples analytical findings were made in breath in 39 cases (3.6%). Most frequently found substances were the following: amphetamine (25 cases) methamphetamine (10 cases), THC (8 cases), cocaine (4 cases), benzoylecgonine (2 cases) and diazepam (2 cases). In conclusion, a fully validated and robust screening method suitable for the routine measurement of drugs of abuse in exhaled breath with a simple procedure for specimen collection and sample preparation was successfully developed.
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ethylene, ≥99.5%
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Oxazepam
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonia solution, 2.0 M in methanol
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol