Merck
Phytochemicals preservation in strawberry as affected by pH modulation.
Food chemistry (2014-10-13)
Ana Oliveira, Maria Helena Gomes, Elisabete M C Alexandre, Fátima Poças, Domingos P F Almeida, Manuela Pintado
ABSTRACT
Strawberries purées are incorporated in foods and subjected to pH modulation according to the expected final food matrix. The effect of pH on strawberry polyphenols stored at 4 and 23 °C for 90 days was evaluated. Total antioxidant activity and total phenolics content were only affected by time according to a first order model. The pH 4.5 induced higher decrease in (-)-epigallocatechin gallate (71% and 79%) and quercetin-3-glucoside (29% and 36%), for both storage temperatures. For pH 2.5 and 3.0, ellagic acid increased 84% for 4 °C and 185% for 23 °C. Anthocyanins concentration changes along storage were well described by first order model. The pH value of 2.5 presented the lower kinetic constant rate where cyanidin-3-glucoside, pelargonidin-3-glucoside and pelargonidin-3-rutinoside had a k=0.04, 0.05 and 0.03 day(-1). Lower storage temperature (4 °C) and lower pH (2.5) were the best condition for the preservation of polyphenols in pasteurized strawberry during a 90-day storage period.
Product Description
Supelco
Potassium Sorbate, analytical standard
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Potassium persulfate, 99.99% trace metals basis
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Supelco
Natamycin, VETRANAL®, analytical standard
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Sodium phosphate dibasic, purum p.a., anhydrous, ≥98.0% (T)
Sigma-Aldrich
Potassium sorbate, purum p.a., ≥99.0% (NT)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Pimaricin, from Streptomyces chattanoogensis, ≥95% (HPLC)
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Supelco
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC
Supelco
(+)-Catechin, analytical standard
Supelco
Gallic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Potassium sorbate, United States Pharmacopeia (USP) Reference Standard
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
USP
Citric acid, United States Pharmacopeia (USP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture