Skip to Content
Merck
All Photos(1)

Key Documents

91215

Supelco

Gallic acid

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

3,4,5-Trihydroxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)3C6H2CO2H
CAS Number:
Molecular Weight:
170.12
Beilstein:
2050274
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage condition

under inert gas (Argon)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

251 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

shipped in

wet ice

storage temp.

2-8°C

SMILES string

OC(=O)c1cc(O)c(O)c(O)c1

InChI

1S/C7H6O5/c8-4-1-3(7(11)12)2-5(9)6(4)10/h1-2,8-10H,(H,11,12)

InChI key

LNTHITQWFMADLM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Gallic acid has been used as a reference standard in the high performance liquid chromatography coupled triple quadrupole mass spectrometric (HPLC/QqQ MS) analysis of gallic acid in rat medicated plural serum containing Guizhi Fuling Wan, a chinese herbal formula used for the treatment of ovarian cancer.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Gallic acid is a water soluble phenolic acid present in grapes and in the leaves of many plants. Gallic acid esters, such as tannins, catechin gallates and aliphatic gallates are potent antioxidants in vitro. However, gallic acid itself also appears to have antioxidant, anticarcinogenic and antiangiogenic activity in vitro.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: eucalyptus humulus

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guizhi fuling wan, a traditional chinese herbal formula, sensitizes cisplatin-resistant human ovarian cancer cells through inactivation of the PI3K/AKT/mTOR pathway
Han L, et al.
Evidence-Based Complementary and Alternative Medicine : ECAM (2016)
Phytochemical Characterization of Rhus coriaria L. Extracts by Headspace Solid-Phase Micro Extraction Gas Chromatography, Comprehensive Two-Dimensional Liquid Chromatography, and Antioxidant Activity Evaluation
Arena K, et al.
Molecules (Basel), 27(5), 1727-1727 (2022)
Acharya Balkrishna et al.
Pharmaceuticals (Basel, Switzerland), 14(4) (2021-04-04)
Divya-Swasari-Vati (DSV) is a calcium-containing herbal medicine formulated for the symptomatic control of respiratory illnesses observed in the current COVID-19 pandemic. DSV is an Ayurvedic medicine used for the treatment of chronic cough and inflammation. The formulation has shown its
Antonio Serrano et al.
Antioxidants (Basel, Switzerland), 8(8) (2019-08-09)
Nowadays, more consumers demand healthier products. A way to offer such products is to functionalize them using health-promoting bioactive compounds. Meat and meat products are high in essential nutrients; however, their excessive consumption implies a high intake of other substances
Parastoo Zarghami Moghaddam et al.
Turkish journal of pharmaceutical sciences, 18(1), 10-16 (2021-02-27)
Cleome coluteoides, which belongs to the Capparidaceae family, and has been used in folk medicine for a long time. Our research aims to measure the antioxidant, antibacterial, and antifungal activities of C. coluteoides. Various solvents, such as ethyl acetate, methanol

Articles

A complete workflow for estimating the total polyphenol content of apple and banana samples, with gallic acid standard addition curve protocol.

A complete workflow for estimating the total polyphenol content of apple and banana samples, with gallic acid standard addition curve protocol.

A complete workflow for estimating the total polyphenol content of apple and banana samples, with gallic acid standard addition curve protocol.

A complete workflow for estimating the total polyphenol content of apple and banana samples, with gallic acid standard addition curve protocol.

Protocols

HPLC Analysis of Polyphenols in Nero d'Avola Red Wine on Discovery® HS C18 (UV 280 nm)

Protocol for HPLC Analysis of Flavonoids on Ascentis® RP-Amide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service