Skip to Content
Merck
All Photos(4)

Documents

S0876

Sigma-Aldrich

Sodium phosphate dibasic

ReagentPlus®, ≥99.0%

Synonym(s):

sec-Sodium phosphate, Disodium hydrogen phosphate, Disodium phosphate, Sodium hydrogenphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4
CAS Number:
Molecular Weight:
141.96
EC Number:
MDL number:
UNSPSC Code:
12161700
eCl@ss:
38070211
PubChem Substance ID:
NACRES:
NA.25

Quality Level

product line

ReagentPlus®

Assay

≥99.0%

form

powder or crystals

pH

8.9-9.2 (25 °C, 50 g/L in water)

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

solubility

water: soluble 100 mg/mL

SMILES string

[Na+].[Na+].[H]OP([O-])([O-])=O

InChI

1S/2Na.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

BNIILDVGGAEEIG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Sodium phosphate dibasic has been used in the establishment of epithelial cell lines from individual rabbit lenses.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Establishment of epithelial cell lines from individual rabbit lenses
Reddan J R, et al.
Methods in Cell Science : An Official Journal of the Society for In Vitro Biology, 6(2), 57-60 (1980)
Leonard Blaschek et al.
Frontiers in plant science, 11, 109-109 (2020-03-21)
Lignin accumulates in the cell walls of specialized cell types to enable plants to stand upright and conduct water and minerals, withstand abiotic stresses, and defend themselves against pathogens. These functions depend on specific lignin concentrations and subunit composition in
Eva Pigna et al.
EBioMedicine, 40, 717-732 (2019-02-05)
Histone deacetylase 4 (HDAC4) has been proposed as a target for Amyotrophic Lateral Sclerosis (ALS) because it mediates nerve-skeletal muscle interaction and since its expression in skeletal muscle correlates with the severity of the disease. However, our recent studies on
Megan A Rees et al.
Journal of proteome research, 14(1), 120-132 (2014-10-21)
Interactions between a host and a bacterial pathogen are mediated by cross-talk between molecules present on, or secreted by, pathogens and host binding-molecules. Identifying proteins involved at this interface would provide substantial insights into this interaction. Although numerous studies have
Kaisorn L Chaichana et al.
Journal of neuroscience methods, 180(1), 116-125 (2009-05-12)
The discoveries of neural (NSCs) and brain tumor stem cells (BTSCs) in the adult human brain and in brain tumors, respectively, have led to a new era in neuroscience research. These cells represent novel approaches to studying normal phenomena such

Articles

Cathepsin B is a lysosomal cysteine proteinase with broad specificity. This protocol uses Nα–CBZ–Arg–Arg–7–amido–4–methylcoumarin as the substrate for fluorometric detection of Cathepsin B activity.

Cathepsin B is a lysosomal cysteine proteinase with broad specificity. This protocol uses Nα–CBZ–Arg–Arg–7–amido–4–methylcoumarin as the substrate for fluorometric detection of Cathepsin B activity.

Cathepsin B is a lysosomal cysteine proteinase with broad specificity. This protocol uses Nα–CBZ–Arg–Arg–7–amido–4–methylcoumarin as the substrate for fluorometric detection of Cathepsin B activity.

Cathepsin B is a lysosomal cysteine proteinase with broad specificity. This protocol uses Nα–CBZ–Arg–Arg–7–amido–4–methylcoumarin as the substrate for fluorometric detection of Cathepsin B activity.

Protocols

Enzymatic Assay of Plasmin with D-Val-Leu-Lys-p-Nitroanilide Dihydrochloride

Protocol for Enzymatic Assay of Plasmin with D-Val-Leu-Lys-p-Nitroanilide Dihydrochloride

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service