Merck
  • Time- and NADPH-dependent inhibition of cytochrome P450 3A4 by the cyclopentapeptide cilengitide: significance of the guanidine group and accompanying spectral changes.
Time- and NADPH-dependent inhibition of cytochrome P450 3A4 by the cyclopentapeptide cilengitide: significance of the guanidine group and accompanying spectral changes.
Drug metabolism and disposition: the biological fate of chemicals (2014-07-06)
Mirza Bojić, Luca Barbero, Hugues Dolgos, Achim Freisleben, Dieter Gallemann, Simona Riva, F Peter Guengerich
ABSTRACT
Cilengitide is a stable cyclic pentapeptide containing an Arg-Gly-Asp motif responsible for selective binding to αVβ3 and αVβ5 integrins. The candidate drug showed unexpected inhibition of cytochrome P450 (P450) 3A4 at high concentrations, that is, a 15-mM concentration caused attenuation of P450 3A4 activity (depending on the probe substrate): 15-19% direct inhibition, 10-23% time-dependent inhibition (30-minute preincubation), and 54-60% metabolism-dependent inhibition (30-minute preincubation). The inactivation efficiency determined with human liver microsomes was 0.003 ± 0.001 min(-1) mM(-1) and was 0.04 ± 0.01 min(-1) mM(-1) with baculovirus-based microsomes containing recombinant P450 3A4. Neither heme loss nor covalent binding to apoprotein could explain the observed reductions in residual activity. Slowly forming type II difference spectra were observed, with maximum spectral changes after 2 hours. Binding to both reduced and oxidized P450 3A4 was observed, with apparent Kd values of 0.66 μM and 6 μM. The significance of the guanidine group in inhibition was demonstrated using ligand binding spectral changes and inactivation assays with guanidine analogs (debrisoquine, N-acetylarginine-O-methyl ester) and the acetylated ornithine derivative of cilengitide. The observed inhibition could be explained by direct inhibition, plus by formation of stable complexes with both ferric and ferrous forms of heme iron and to some extent by the formation of reactive species capable to react to the protein or heme. Formation of the complex required time and NADPH and is attributed to the guanidino group. Thus, the NADPH-dependent inhibition is considered to be mainly due to the formation of a stable complex rather than the formation of reactive species.
Sigma-Aldrich
Cytochrome P450 human, 1A2 Isozyme Microsomes, with P450 Reductase and cytochrome b5, recombinant, expressed in baculovirus infected insect cells (BTI-TN-5B1-4)
Sigma-Aldrich
Cytochrome P450 human, 1A1 Isozyme Microsomes, with P450 Reductase, recombinant, expressed in baculovirus infected insect cells (BTI-TN-5B1-4)
Sigma-Aldrich
Cytochrome P450 human, 4F3B isozyme microsomes, with P450 Reductase and cytochrome b5, recombinant, expressed in baculovirus infected insect cells (BTI-TN-5B1-4)
Sigma-Aldrich
Cytochrome P450 human, 3A4 isozyme microsomes, with P450 reductase and cytochrome b5, recombinant, expressed in baculovirus infected insect cells (BTI-TN-5B1-4)
Sigma-Aldrich
Cytochrome P450 human, 2E1 isozyme microsomes, with P450 Reductase and cytochrome b5, recombinant, expressed in baculovirus infected insect cells (BTI-TN-5B1-4)
Sigma-Aldrich
Cytochrome P450 human, 1B1 Isozyme Microsomes, with P450 Reductase, recombinant, expressed in baculovirus infected insect cells (BTI-TN-5B1-4)
Supelco
Testosterone, VETRANAL®, analytical standard
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Nifedipine, European Pharmacopoeia (EP) Reference Standard
Supelco
Nifedipine, Pharmaceutical Secondary Standard; Certified Reference Material
Testosterone, European Pharmacopoeia (EP) Reference Standard
SAFC
BIS-TRIS
USP
Nifedipine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Testosterone, ≥98%
SAFC
BIS-TRIS