Skip to Content
Merck

Skip To

1463508

USP

Nifedipine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1,4-Dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylic acid dimethyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C17H18N2O6
CAS Number:
Molecular Weight:
346.33
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
244-598-3
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

nifedipine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

COC(=O)C1=C(C)NC(C)=C(C1c2ccccc2[N+]([O-])=O)C(=O)OC

InChI

1S/C17H18N2O6/c1-9-13(16(20)24-3)15(14(10(2)18-9)17(21)25-4)11-7-5-6-8-12(11)19(22)23/h5-8,15,18H,1-4H3

InChI key

HYIMSNHJOBLJNT-UHFFFAOYSA-N

Gene Information

General description

Find answers to frequently asked questions on USP products in USP FAQs.

Application

Nifedipine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Nifedipine Capsules
  • Nifedipine Extended-Release Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1463224PHR1290N0750000
description

United States Pharmacopeia (USP) Reference Standard

description

United States Pharmacopeia (USP) Reference Standard

description

Pharmaceutical Secondary Standard; Certified Reference Material

description

European Pharmacopoeia (EP) Reference Standard

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

EDQM

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

format

neat

format

neat

format

neat

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

Gene Information

human ... CACNA1C(775), CACNA1D(776), CACNA1F(778), CACNA1S(779)

Gene Information

human ... CACNA1C(775), CACNA1D(776), CACNA1F(778), CACNA1S(779)

Gene Information

human ... CACNA1C(775), CACNA1D(776), CACNA1F(778), CACNA1S(779)

Gene Information

human ... CACNA1C(775), CACNA1D(776), CACNA1F(778), CACNA1S(779)

agency

USP

agency

USP

agency

traceable to Ph. Eur. N0750000, traceable to USP 1463508

agency

-


Still not finding the right product?

Explore all of our products under Nifedipine


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters