Merck
  • Copolyesters made from 1,4-butanediol, sebacic acid, and D-glucose by melt and enzymatic polycondensation.
Copolyesters made from 1,4-butanediol, sebacic acid, and D-glucose by melt and enzymatic polycondensation.
Biomacromolecules (2015-01-27)
Cristina Japu, Antxon Martínez de Ilarduya, Abdelilah Alla, Yi Jiang, Katja Loos, Sebastián Muñoz-Guerra
ABSTRACT
Biotechnologically accessible 1,4-butanediol and vegetal oil-based diethyl sebacate were copolymerized with bicyclic acetalized D-glucose derivatives (Glux) by polycondensation both in the melt at high temperature and in solution at mild temperature mediated by polymer-supported Candida antarctica lipase B (CALB). Two series of random copolyesters (PB(x)Glux(y)Seb and PBSeb(x)Glux(y)) were prepared differing in which d-glucose derivative (Glux diol or Glux diester) was used as comonomer. The three parent homopolyesters PBSeb, PBGlux, and PGluxSeb were prepared as well. Both methods were found to be effective for polymerization although significant higher molecular weights were achieved by melt polycondensation. The thermal properties displayed by the copolyesters were largely dependent on composition and also on the functionality of the replacing Glux unit. The thermal stability of PBSeb was retained or even slightly increased after copolymerization with Glux, whereas crystallinity and melting temperature were largely depressed. On the contrary, the glass-transition temperature noticeably increased with the content in Glux units. PGluxSeb distinguished in displaying both T(g) and T(m) higher than PBSeb because a different crystal structure is adopted by this homopolyester. The hydrolytic degradability of PBSeb in water was enhanced by copolymerization, in particular, when biodegradation was assisted by lipases.
Product Description
Supelco
1,1,1,3,3,3-Hexafluoro-2-propanol, for GC derivatization, LiChropur, ≥99.8%
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
1,1,1,3,3,3-Hexafluoro-2-propanol, ≥99%
Sigma-Aldrich
Dibutyltin(IV) oxide, 98%
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), for molecular biology
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sorbitol, European Pharmacopoeia (EP) Reference Standard
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sorbitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Diethyl sebacate, 98%
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
1,4-Butanediol, ReagentPlus®, 99%
Sigma-Aldrich
Sodium trifluoroacetate, 98%
Sigma-Aldrich
Diethyl sebacate, ≥98%, FCC, FG