Merck
  • Systematic evaluation of commercially available ultra-high performance liquid chromatography columns for drug metabolite profiling: optimization of chromatographic peak capacity.
Systematic evaluation of commercially available ultra-high performance liquid chromatography columns for drug metabolite profiling: optimization of chromatographic peak capacity.
Journal of chromatography. A (2014-12-02)
Anne-Charlotte Dubbelman, Filip Cuyckens, Lieve Dillen, Gerhard Gross, Thomas Hankemeier, Rob J Vreeken
ABSTRACT
The present study investigated the practical use of modern ultra-high performance liquid chromatography (UHPLC) separation techniques for drug metabolite profiling, aiming to develop a widely applicable, high-throughput, easy-to-use chromatographic method, with a high chromatographic resolution to accommodate simultaneous qualitative and quantitative analysis of small-molecule drugs and metabolites in biological matrices. To this end, first the UHPLC system volume and variance were evaluated. Then, a mixture of 17 drugs and various metabolites (molecular mass of 151-749Da, logP of -1.04 to 6.7), was injected on six sub-2μm particle columns. Five newest generation core shell technology columns were compared and tested against one column packed with porous particles. Two aqueous (pH 2.7 and 6.8) and two organic mobile phases were evaluated, first with the same flow and temperature and subsequently at each column's individual limit of temperature and pressure. The results demonstrated that pre-column dead volume had negligible influence on the peak capacity and shape. In contrast, a decrease in post-column volume of 57% resulted in a substantial (47%) increase in median peak capacity and significantly improved peak shape. When the various combinations of stationary and mobile phases were used at the same flow rate (0.5mL/min) and temperature (45°C), limited differences were observed between the median peak capacities, with a maximum of 26%. At higher flow though (up to 0.9mL/min), a maximum difference of almost 40% in median peak capacity was found between columns. The finally selected combination of solid-core particle column and mobile phase composition was chosen for its selectivity, peak capacity, wide applicability and peak shape. The developed method was applied to rat hepatocyte samples incubated with the drug buspirone and demonstrated to provide a similar chromatographic resolution, but a 6 times higher signal-to-noise ratio than a more traditional UHPLC metabolite profiling method using a fully porous particle packed column, within one third of the analysis time. In conclusion, a widely applicable, selective and fast chromatographic method was developed that can be applied to perform drug metabolite profiling in the timeframe of a quantitative analysis. It is envisioned that this method will in future be used for simultaneous qualitative and quantitative analysis and can therefore be considered a first important step in the Quan/Qual workflow.
Sigma-Aldrich
Darunavir, ≥98% (HPLC)
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Risperidone, ≥98% (HPLC), powder
Loperamide hydrochloride, European Pharmacopoeia (EP) Reference Standard
Norethisterone, European Pharmacopoeia (EP) Reference Standard
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Supelco
Loperamide hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
4-Hydroxymidazolam
Supelco
Loperamide hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Galanthamine hydrobromide from Lycoris sp., ≥94% (HPLC)
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Prednisone, ≥98%
Sigma-Aldrich
19-Norethindrone, ≥98%, powder
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Risperidone, European Pharmacopoeia (EP) Reference Standard
Prednisone, European Pharmacopoeia (EP) Reference Standard
USP
Loperamide hydrochloride, United States Pharmacopeia (USP) Reference Standard