Skip to Content
Merck

34014

Loperamide hydrochloride

VETRANAL®, analytical standard

Synonym(s):

4-(p-Chlorophenyl)-4-hydroxy-N,N-dimethyl-α,α-diphenyl-1-piperidinebutyramide hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C29H33ClN2O2 · HCl
CAS Number:
Molecular Weight:
513.50
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
252-082-4
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CN(C)C(=O)C(CCN1CCC(O)(CC1)c2ccc(Cl)cc2)(c3ccccc3)c4ccccc4

InChI

1S/C29H33ClN2O2.ClH/c1-31(2)27(33)29(24-9-5-3-6-10-24,25-11-7-4-8-12-25)19-22-32-20-17-28(34,18-21-32)23-13-15-26(30)16-14-23;/h3-16,34H,17-22H2,1-2H3;1H

InChI key

PGYPOBZJRVSMDS-UHFFFAOYSA-N

Gene Information

human ... OPRM1(4988)

General description

Loperamide is an antidiarrheal agent used in controlling acute nonspecific diarrhea and chronic diarrhea associated with inflammatory bowel diseases.

Application

Loperamide hydrochloride may be used as an analytical reference standard for the quantification of the analyte in biological fluids using various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Loperamide hydrochloride


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Quantitation of loperamide and N-demethyl-loperamide in human plasma using electrospray ionization with selected reaction ion monitoring liquid chromatography-mass spectrometry.
He H, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 744(323-331), 2-2 (2000)
LC-MS determination and bioavailability study of loperamide hydrochloride after oral administration of loperamide capsule in human volunteers.
Yu H-J, et al.
Journal of Pharmaceutical and Biomedical Analysis, 36(2), 421-427 (2004)
James Church et al.
Diseases of the colon and rectum, 56(1), 120-125 (2012-12-12)
Coating of the cecum with sticky bile causes a problem with inspection of the mucosa during colonoscopy. This study aimed to see whether 4 mg of loperamide taken after colonoscopy preparation would delay the passage of bile and improve the



Global Trade Item Number

SKUGTIN
34014-100MG-R04061834243953