Merck
  • Structure-activity relationships of a novel pyranopyridine series of Gram-negative bacterial efflux pump inhibitors.
Structure-activity relationships of a novel pyranopyridine series of Gram-negative bacterial efflux pump inhibitors.
Bioorganic & medicinal chemistry (2015-03-31)
Son T Nguyen, Steven M Kwasny, Xiaoyuan Ding, Steven C Cardinale, Courtney T McCarthy, Hong-Suk Kim, Hiroshi Nikaido, Norton P Peet, John D Williams, Terry L Bowlin, Timothy J Opperman
ABSTRACT
Recently we described a novel pyranopyridine inhibitor (MBX2319) of RND-type efflux pumps of the Enterobacteriaceae. MBX2319 (3,3-dimethyl-5-cyano-8-morpholino-6-(phenethylthio)-3,4-dihydro-1H-pyrano[3,4-c]pyridine) is structurally distinct from other known Gram-negative efflux pump inhibitors (EPIs), such as 1-(1-naphthylmethyl)-piperazine (NMP), phenylalanylarginine-β-naphthylamide (PAβN), D13-9001, and the pyridopyrimidine derivatives. Here, we report the synthesis and biological evaluation of 60 new analogs of MBX2319 that were designed to probe the structure activity relationships (SARs) of the pyranopyridine scaffold. The results of these studies produced a molecular activity map of the scaffold, which identifies regions that are critical to efflux inhibitory activities and those that can be modified to improve potency, metabolic stability and solubility. Several compounds, such as 22d-f, 22i and 22k, are significantly more effective than MBX2319 at potentiating the antibacterial activity of levofloxacin and piperacillin against Escherichia coli.
Sigma-Aldrich
Indole, ≥99%, FG
Sigma-Aldrich
1-Methylhomopiperazine, 97%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
1H-Pyrazole-1-carboxamidine hydrochloride, 99%
Sigma-Aldrich
Indole, ≥99%
Sigma-Aldrich
Pyrrolidine, 99%
Sigma-Aldrich
Dimethylamine, anhydrous, ≥99%
Sigma-Aldrich
Thiomorpholine, 98%
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥98%
Sigma-Aldrich
Methyl-β-D-thiogalactoside
USP
Levofloxacin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Pyrrolidine, FG
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Pyrrolidine, ≥99.0%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
3-Chlorophenethyl bromide, 97%
Sigma-Aldrich
3-Bromophenethyl bromide, 97%
Supelco
Carbon disulfide solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
4-Methoxyphenethyl bromide, 97%
Sigma-Aldrich
Trimethylgallium, packaged for use in deposition systems
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Supelco
Methanol, analytical standard
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Pyrrolidine, ≥99.5%, purified by redistillation
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
1-(2-Bromoethyl)-3-fluorobenzene, AldrichCPR
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%