Skip to Content
Merck
All Photos(1)

Key Documents

83240

Sigma-Aldrich

Pyrrolidine

≥99.0%

Synonym(s):

Tetrahydropyrrole, Tetramethyleneimine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9N
CAS Number:
Molecular Weight:
71.12
Beilstein:
102395
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

2.45 (vs air)

Quality Level

vapor pressure

128 mmHg ( 39 °C)
49 mmHg ( 20 °C)

Assay

≥99.0% (GC)
≥99.0%

form

liquid

autoignition temp.

653 °F

expl. lim.

10.6 %

impurities

≤0.5% water

refractive index

n20/D 1.443 (lit.)
n20/D 1.443

density

0.852 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.4 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

C1CCNC1

InChI

1S/C4H9N/c1-2-4-5-3-1/h5H,1-4H2

InChI key

RWRDLPDLKQPQOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Pyrrolidine can be used as:
  • A base for the chemoselective reduction of ketones to alcohol in the presence of decaborane and cerium(III) chloride heptahydrate in methanol.[1]
  • A reactant with benzaldehyde and β-naphthol for the synthesis of betti base using non-ionic surfactant (Triton X-100) as a catalyst.[2]
  • A catalyst in the synthesis of aldimines from aldehydes and primary amines.[3]
  • A modifier for aluminum hydride reagent, which is used as a selective reducing agent to transform N-benzyl-4-ethoxycarbonylpiperidine into N-benzyl-4-formylpiperidine.[4]
  • As a solvent/base in palladium-catalyzed Sonagashira coupling reactions.[5]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

37.4 °F - closed cup

Flash Point(C)

3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid, easy copper-free Sonogashira couplings using aryl iodides and activated aryl bromides
Leadbeater NE and Tominack BJ
Tetrahedron Letters, 44(48), 8653-8656 (2003)
Large scale synthesis of N-benzyl-4-formylpiperidine through partial reduction of esters using aluminum hydride reagents modified with pyrrolidine
Abe T, et al.
Tetrahedron, 57(14), 2701-2710 (2001)
Reduction of ketones to alcohols using a decaborane/pyrrolidine/cerium (III) chloride system in methanol
Bae JW, et al.
Tetrahedron Letters, 42(11), 2137-2139 (2001)
Non-ionic surfactant catalyzed synthesis of Betti base in water
Kumar A, et al.
Tetrahedron Letters, 51(12), 1582-1584 (2010)
A general aminocatalytic method for the synthesis of aldimines
Morales S, et al.
Journal of the American Chemical Society, 136(3), 1082-1089 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service