Merck
  • Astragaloside IV inhibits renal tubulointerstitial fibrosis by blocking TGF-β/Smad signaling pathway in vivo and in vitro.
Astragaloside IV inhibits renal tubulointerstitial fibrosis by blocking TGF-β/Smad signaling pathway in vivo and in vitro.
Experimental biology and medicine (Maywood, N.J.) (2014-06-01)
Li Wang, Yang-Feng Chi, Ze-Ting Yuan, Wen-Chao Zhou, Pei-Hao Yin, Xue-Mei Zhang, Wen Peng, Hui Cai
ABSTRACT
Astragaloside IV (AS-IV) is a major active ingredient from Radix astragali, which has been considered as a renoprotective agent; however, its molecular mechanisms are unclear. Thus, we designed to investigate the renoprotective effects and mechanisms of AS-IV in rat model of renal fibrosis induced by unilateral ureteral obstruction (UUO) in vivo and TGF-β1-stimulated rat renal fibroblasts (NRK-49F) in vitro. Sprague-Dawley rats were randomly divided into six groups: sham operation, UUO, UUO/AS-IV (3.3, 10, 33 mg·kg(-1)·d(-1)), and UUO/enalapril (4 mg·kg(-1)·d(-1)). Renal function, tubulointerstitial damage index score, extracellular matrix (ECM) deposition, and the expressions of TGF-β1, connective tissue growth factor (CTGF), α-SMA, fibronectin, collagen I, III, Smad2/3, phosphorylated-Smad2/3, and Smad7 were measured. In addition, the expressions of CTGF, α-SMA, fibronectin, collagen I, III, Smad2/3, phosphorylated-Smad2/3, and Smad7 were measured in TGF-β1-stiumlated NRK-49F cell line. AS-IV significantly decreased UUO-induced renal fibrosis and functional impairment, which are associated with inhibition of TGF-β1, CTGF, α-SMA, and collagen matrix expression, and a decrease in serum creatinine and urea nitrogen. The renoprotective effects of AS-IV on fibrosis were associated with up-regulation of Smad7, thereby blocking up-regulations of TGF-β1, CTGF, and α-SMA, and activation of phosphorylated-Smad2/3. These effects were further conformed in NRK-49F cell line stimulated by TGF-β1. Moreover, knockdown of Smad7 gene in NRK-49F cells was able to prevent AS-IV-induced inhibition to Smad2/3 signaling activation, expression of CTGF, α-SMA, and ECM proteins in response to TGF-β1. Renal tubulointerstitial fibrosis was attenuated by treatment with AS-IV, which was closely related to induction of Smad7, thereby inhibiting TGF-β/Smad signaling.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Millipore
Astragaloside IV, >98.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Astragaloside IV, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%