Merck
  • Multiresidue method for pesticides in drinking water using a graphitized carbon black cartridge extraction and liquid chromatographic analysis.
Multiresidue method for pesticides in drinking water using a graphitized carbon black cartridge extraction and liquid chromatographic analysis.
Analytical chemistry (1991-03-15)
A Di Corcia, M Marchetti
ABSTRACT
A general liquid-solid extraction procedure for the isolation of pesticides from groundwater and drinking water for high-performance liquid chromatography (HPLC) is presented. This simple and rapid procedure involved passing a 2-L sample through a 250-mg graphitized carbon black (Carbopack B) cartridge at a flow rate of 150-160 mL/min. By taking advantage of the presence of positively charged active centers on the Carbopack B surface, a stepwise elution system allowed the complete separation of base-neutral pesticides from acidic ones. After partial solvent removal, the components in the two fractions were separated and quantified by gradient elution, reversed-phase HPLC with ultraviolet (UV) detection. The performance of the Carbopack cartridge was compared with that of a 500-mg C-18 bonded silica cartridge. With the Carbopack cartridge, the grand mean measurement accuracy of the 35 pesticides considered was 95%. With the C-18 cartridge, the grand mean measurement accuracy of the analytes was 76%. Compared to the C-18 cartridge, additional advantages of using a Carbopack cartridge are that the extraction procedure is about 7 times shorter, no pH adjustment of the environmental sample is necessary for trapping acidic compounds, and one cartridge instead of two suffices to extract base-neutral and acidic pesticides, making the Carbopack cartridge more adaptable than the C-18 one for field use. The detection limits by this method of all the pesticides considered were between 0.003 and 0.07 micrograms/L.
Product Description
Supelco
Carbopack Adsorbent, matrix Carbopack C, 80-100 mesh, bottle of 10 g
Supelco
Carbopack Adsorbent, matrix Carbopack X, 40-60 mesh, bottle of 10 g
Supelco
Carbopack Adsorbent, matrix Carbopack X, 120-400 mesh, bottle of 50 g
Supelco
Metal Packed GC Column (fits PerkinElmer®), phase 3% SP-1500, matrix 80/120 Carbopack B support, L × O.D. × I.D. 10.0 ft (3.0 m) × 1/8 in. × 2.1 mm
Supelco
Carbopack Adsorbent, matrix Carbopack C, 60-80 mesh, bottle of 10 g
Supelco
Carbopack B, stainless steel TD tube, O.D. × L 1/4 × 3 1/2, unconditioned, pkg of 10 ea
Supelco
Carbopack B, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., unconditioned, pkg of 10 ea, matrix Carbopack
Supelco
Carbopack B, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
Carbopack B, O.D. × L 1/4 in. × 3 1/2 in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
Supelco
Carbopack Adsorbent, matrix Carbopack X, 60-80 mesh, bottle of 10 g
Supelco
FLM Carbopack X Deactivated Stainless Steel TD Tube, preconditioned, pkg of 10 tubes, O.D. × L 1/4 in. (6.35 mm) × 3 1/2 in. (89 mm)
Supelco
Small Particle GCB Adsorbent, matrix Graphitized Carbon, <200 nm, bottle of 50 g
Supelco
Metal Packed GC Column (General Configuration), phase 5% Krytox 143AC, matrix 60/80 Carbopack B support, L × O.D. × I.D. 40.0 ft (12.2 m) × 1/8 in. × 1.7 mm
Supelco
Carbopack X, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
Small Particle GCB Adsorbent, matrix Mesoporous Carbon, 45 μm, bottle of 50 g
Supelco
Carbopack Adsorbent, matrix Carbopack B, 60-80 mesh, bottle of 10 g