30135-U
Carbopack™ B Thermal Desorption Tube
stainless steel, O.D. x I.D. x L 1/4 × 3 1/2 , unconditioned, pkg of 10 ea
Synonym(s):
Carbon adsorbent, TD Tube
About This Item
Recommended Products
product name
Carbopack™ B, stainless steel TD tube, O.D. × L 1/4 × 3 1/2, unconditioned, pkg of 10 ea
material
stainless steel TD tube
product line
Carbopack™
feature
unconditioned
packaging
pkg of 10 ea
greener alternative product characteristics
Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
technique(s)
active air sampling: suitable
O.D. × L
1/4 × 3 1/2
surface area
~100 m2/g
matrix
Carbopack™ graphitized carbon black (GCB)
suitability
suitable for (For sampling a wide range of VOCs)
application(s)
air monitoring
environmental
industrial hygiene
compatibility
PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments
greener alternative category
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service