Merck
  • Layer-by-layer construction of protein architectures through avidin-biotin and lectin-sugar interactions for biosensor applications.
Layer-by-layer construction of protein architectures through avidin-biotin and lectin-sugar interactions for biosensor applications.
Analytical and bioanalytical chemistry (2011-08-26)
Shigehiro Takahashi, Katsuhiko Sato, Jun-ichi Anzai
ABSTRACT
In this review, the preparation and properties of protein architectures constructed by layer-by-layer (LbL) deposition through avidin-biotin and concanavalin A (Con A)-sugar interactions are discussed in relation to their use for optical and electrochemical biosensors. LbL films can be constructed through the alternate deposition of avidin and biotin-labeled enzymes on the surfaces of optical probes and electrodes. The enzymes retain their catalytic activity, resulting in the formation of optical and electrochemical biosensors. Alternatively, Con A can be used to construct enzyme-containing LbL films and microcapsules using sugar-labeled enzymes. Some enzymes such as glucose oxidase and horseradish peroxidase can be used for this purpose without labeling with sugar, because these enzymes contain intrinsic hydrocarbon chains on their molecular surfaces. The Con A/enzyme LbL architectures were successfully used to develop biosensors sensitive to specific substrates of the enzyme. In addition, Con A-based films can be used for the optical and electrochemical detection of sugars.
Product Description
Supelco
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Biotin, meets USP testing specifications
Biotin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
USP
Biotin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
Supelco
Biotin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Biotin, ≥99.0% (T)
SAFC
BIOTIN