Merck
Learning experience in endodontics: Brazilian students' perceptions.
Journal of dental education (2013-05-10)
Marilia O S Seijo, Efigênia F Ferreira, Antônio P Ribeiro Sobrinho, Saul M Paiva, Renata C Martins
ABSTRACT
Including students' perceptions in the educational process is considered a key component in monitoring the quality of academic programs. This study aimed to evaluate the concept of one's learning experience in endodontic teaching from the perspective of a group of Brazilian students. A total of 126 self-administered, structured questionnaires were distributed to undergraduate dental students enrolled in endodontics courses during the second semester of the 2009 academic year. The questionnaires were administered during final examinations and focused on students' opinions concerning learning during endodontic treatments, time spent during endodontic treatments, difficulties found during endodontic treatments, quality of endodontic treatments performed, characteristics of the technique employed, and suggestions to improve endodontic teaching. Ninety-one percent of the questionnaires were returned for evaluation. The obtained answers were discussed and analyzed, thereby generating quantitative and qualitative data showing students' perceptions of their experiences in endodontics courses. The main points that can affect the teaching of endodontics, according to the undergraduate students, included patients' absences and delays, selection of patients, preclinical and clinical training, difficulties found, type of technique employed, and teachers' orientation during endodontic treatment. The students' perceptions provided valuable information about the development of the course and the teacher-student relationship, together with the added intention of enhancing the teaching of endodontics as well as other courses.
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2400, slurry, in H2O, active catalyst
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 3202, slurry, in H2O, active catalyst
Sigma-Aldrich
Nickel, foil, thickness 0.25 mm, 99.995% trace metals basis
Sigma-Aldrich
Nickel, powder, <1 μm, 99.8% trace metals basis
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 4200, slurry, in H2O, active catalyst
Sigma-Aldrich
Nickel, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2800, slurry, in H2O, active catalyst
Sigma-Aldrich
Nickel, foil, thickness 0.125 mm, ≥99.9%
Sigma-Aldrich
Nickel, wire, diam. 0.25 mm, ≥99.9%
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis