Skip to Content
Merck
All Photos(1)

Key Documents

268283

Sigma-Aldrich

Nickel

powder, <1 μm, 99.8% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ni
CAS Number:
Molecular Weight:
58.69
EC Number:
MDL number:
UNSPSC Code:
12141729
PubChem Substance ID:
NACRES:
NA.23

Assay

99.8% trace metals basis

form

powder

resistivity

6.97 μΩ-cm, 20°C

particle size

<1 μm

bp

2732 °C (lit.)

mp

1453 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Ni]

InChI

1S/Ni

InChI key

PXHVJJICTQNCMI-UHFFFAOYSA-N

Testing Method

Average particle size determined by Fisher sub-sieve sizer

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Qing Qi et al.
ACS applied materials & interfaces, 12(32), 36568-36577 (2020-07-21)
It is well-known that attractive electromagnetic interference (EMI) shielding performance depends on functional (e.g., electrical and magnetic) fillers and structural designs. This paper presents a novel three-layered sandwich structure of poly(vinylidene fluoride) (PVDF)-based nanocomposites, consisting of graphene nanoplatelets (GNP), nickel
Ornella Valenti et al.
Cerebral cortex (New York, N.Y. : 1991), 19(3), 658-674 (2008-07-18)
The prefrontal cortex receives multiple inputs from the hippocampal complex, which are thought to drive memory-guided behavior. Moreover, dysfunctions of both regions have been repeatedly associated with several psychiatric disorders. Therefore, understanding the interconnections and modulatory interactions between these regions
S M Marchan et al.
The European journal of prosthodontics and restorative dentistry, 21(1), 24-28 (2013-05-21)
Posterior resin bonded cast metal restorations (adhesive onlays) were used in a variety of clinical scenarios including: management of tooth wear and cracked tooth, as retainers for fixed bridge work, for correction of the occlusal plane and in providing cuspal
Brenda L Bloodgood et al.
PLoS biology, 7(9), e1000190-e1000190 (2009-09-16)
Excitatory synapses on mammalian principal neurons are typically formed onto dendritic spines, which consist of a bulbous head separated from the parent dendrite by a thin neck. Although activation of voltage-gated channels in the spine and stimulus-evoked constriction of the
Picture of the month. Allergic contact dermatitis to nickel-containing dental work.
Emma F Johnson et al.
JAMA pediatrics, 167(6), 581-582 (2013-06-05)

Articles

Permanent magnets are an essential technology for energy conversion. Motors and generators are used to convert energy between electrical and mechanical forms.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service