Merck
  • Overexpression, purification and crystallization of PhzA, the first enzyme of the phenazine biosynthesis pathway of Pseudomonas fluorescens 2-79.
Overexpression, purification and crystallization of PhzA, the first enzyme of the phenazine biosynthesis pathway of Pseudomonas fluorescens 2-79.
Acta crystallographica. Section D, Biological crystallography (2004-05-26)
Ekta G Ahuja, Dimitri V Mavrodi, Linda S Thomashow, Wulf Blankenfeldt
ABSTRACT
Phenazines are broad-spectrum antibiotic metabolites produced by organisms such as Pseudomonas and Streptomyces. Phenazines have been shown to enhance microbial competitiveness and the pathogenic potential of the organisms that synthesize them. PhzA (163 residues, approximate molecular weight 18.7 kDa) is the product of the first of seven genes of the operon responsible for phenazine biosynthesis in P. fluorescens 2-79. This enzyme is thought to catalyse one of the final steps in the formation of phenazine-1-carboxylic acid, the end product of phenazine biosynthesis in P. fluorescens 2-79. Here, the purification and crystallization of recombinant PhzA are reported. Crystals diffracting to 2.1 angstroms were obtained using 1.6 M magnesium sulfate and 2-morpholinoethanesulfonic acid monohydrate (MES) buffer pH 5.2-5.6. Crystals of both native and seleno-L-methionine-labelled protein belong to the orthorhombic space group P2(1)2(1)2(1), with unit-cell parameters a = 66.8, b = 75.3, c = 84.5 angstroms. The asymmetric unit contains one dimer of PhzA.
Sigma-Aldrich
MES hydrate, BioPerformance Certified, suitable for cell culture, ≥99.5%
Sigma-Aldrich
MES hydrate, ≥99.5% (titration)
Sigma-Aldrich
MES hydrate, ≥99.5% (titration), pH 2.5-4.0 (0.5 M in H2O), BioXtra
Sigma-Aldrich
MES hydrate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
MES solution, BioUltra, Molecular Biology, 0.5 M in H2O
Sigma-Aldrich
MES sodium salt, ≥99% (titration)
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
MES sodium salt, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
MES sodium salt, BioReagent, ≥99% (titration), crystalline
SAFC
MES sodium salt