Skip to Content
Merck

Key Documents

Skip To

76039

MES solution

BioUltra, Molecular Biology, 0.5 M in H2O

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C6H13NO4S
CAS Number:
Molecular Weight:
195.24
NACRES:
NA.25
PubChem Substance ID:
eCl@ss:
32129211
UNSPSC Code:
12161700
MDL number:
Beilstein/REAXYS Number:
6350956
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Segment

product line

BioUltra

form

liquid

concentration

0.5 M in H2O

pH

2.5-4.0 (25 °C in neat)

anion traces

chloride (Cl-): ≤10 mg/kg, sulfate (SO42-): ≤10 mg/kg

cation traces

Al: ≤1 mg/kg, Ba: ≤1 mg/kg, Bi: ≤1 mg/kg, Ca: ≤5 mg/kg, Cd: ≤1 mg/kg, Co: ≤1 mg/kg, Cr: ≤1 mg/kg, Cu: ≤1 mg/kg, Fe: ≤1 mg/kg, K: ≤20 mg/kg, Li: ≤1 mg/kg, Mg: ≤1 mg/kg, Mn: ≤1 mg/kg, Mo: ≤1 mg/kg, Na: ≤20 mg/kg, Ni: ≤1 mg/kg, Pb: ≤1 mg/kg, Sr: ≤1 mg/kg, Zn: ≤1 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: 0.01, λ: 280 nm Amax: 0.01

SMILES string

OS(=O)(=O)CCN1CCOCC1

InChI

1S/C6H13NO4S/c8-12(9,10)6-3-7-1-4-11-5-2-7/h1-6H2,(H,8,9,10)

InChI key

SXGZJKUKBWWHRA-UHFFFAOYSA-N

Application


  • Nanometric determination of the thickness of aqueous samples for accurate molar absorption coefficients of water-soluble molecules in the mid-infrared region.: This study utilizes MES solution as a buffering agent to achieve precise thickness measurements of aqueous samples. The accurate molar absorption coefficients obtained enhance the reliability of spectroscopic analyses in biochemical research (Gutierrez-Salazar & Lorenz-Fonfria, 2024).

  • pH-Responsive Nanogels Generated by Polymerization-Induced Self-Assembly of a Succinate-Functional Monomer.: This research employs MES solution to maintain the desired pH environment during the synthesis of pH-responsive nanogels. The study highlights the potential applications of these nanogels in drug delivery systems (Du & Fielding, 2024).



Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M36719330293283
description

BioUltra, Molecular Biology, 0.5 M in H2O

description

low moisture content, ≥99% (titration)

description

BioUltra, Molecular Biology, pH 7.4

description

BioUltra, Molecular Biology, pH 8.0

grade

Molecular Biology

grade

-

grade

Molecular Biology

grade

Molecular Biology

concentration

0.5 M in H2O

concentration

-

concentration

-

concentration

-

Quality Level

200

Quality Level

400

Quality Level

100

Quality Level

200

product line

BioUltra

product line

-

product line

BioUltra

product line

BioUltra

form

liquid

form

powder

form

liquid

form

liquid

pH

2.5-4.0 (25 °C in neat)

pH

2.5-4 (25 °C, 97.6 g/L)

pH

7.4±0.2 (25 °C)

pH

8.0±0.2 (25 °C)


Storage Class

10 - Combustible liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters