Merck
  • Integration of a contactless conductivity detector into a commercial capillary cassette. Detection of inorganic cations and catecholamines.
Integration of a contactless conductivity detector into a commercial capillary cassette. Detection of inorganic cations and catecholamines.
Journal of chromatography. A (2003-04-11)
Pasi S Vuorinen, Matti Jussila, Heli Sirén, Sami Palonen, Marja-Liisa Riekkola
ABSTRACT
A contactless conductivity detector integrated into the capillary cassette of Agilent (3D)CE equipment is described. The detector is user-friendly, compact and easily modified. The UV detector of the (3D)CE equipment is available parallel with the contactless conductivity detector increasing the detection power. Two electrolyte solutions, 2-(N-morpholino)ethanesulfonic acid-histidine solution (20 mM, pH 6.0) and ammonium acetate (10 mM, pH 4.0), were used as the separation media for inorganic cations and organic catecholamines, respectively. The detection limit for all metal cations except barium was under 0.5 mg/l, and that for four catecholamines was ca. 10 mg/l. This last value was the same order of magnitude as achieved with parallel UV detection.
Sigma-Aldrich
MES hydrate, BioPerformance Certified, suitable for cell culture, ≥99.5%
Sigma-Aldrich
MES hydrate, ≥99.5% (titration)
Sigma-Aldrich
MES hydrate, ≥99.5% (titration), pH 2.5-4.0 (0.5 M in H2O), BioXtra
Sigma-Aldrich
MES hydrate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
MES solution, BioUltra, Molecular Biology, 0.5 M in H2O
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
MES sodium salt, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
MES sodium salt, BioReagent, ≥99% (titration), crystalline
Sigma-Aldrich
MES sodium salt, ≥99% (titration)
SAFC
MES sodium salt