Merck
  • Genome sequence of Pseudomonas sp. Strain S9, an extracellular arylsulfatase-producing bacterium isolated from Mangrove Soil.
Genome sequence of Pseudomonas sp. Strain S9, an extracellular arylsulfatase-producing bacterium isolated from Mangrove Soil.
Journal of bacteriology (2011-05-31)
Mengxian Long, Lingwei Ruan, Ziniu Yu, Xun Xu
ABSTRACT
Pseudomonas sp. strain S9 was originally isolated from mangrove soil in Xiamen, China. It is an aerobic bacterium which shows extracellular arylsulfatase activity. Here, we describe the 4.8-Mb draft genome sequence of Pseudomonas sp. S9, which exhibits novel cysteine-type sulfatases.
Sigma-Aldrich
Sulfatase from Aerobacter aerogenes, Type VI, buffered aqueous glycerol solution, 2-5 units/mg protein (biuret), 10-20 units/mL
Sigma-Aldrich
Sulfatase from Helix pomatia, Type H-2, aqueous solution, ≥2,000 units/mL
Sigma-Aldrich
Sulfatase from Helix pomatia, Type H-1, sulfatase ≥10,000 units/g solid
Sigma-Aldrich
Sulfatase from Patella vulgata (keyhole limpet), Type IV, essentially salt-free, lyophilized powder, ≥10 units/mg solid
Sigma-Aldrich
Sulfatase from abalone entrails, Type VIII, lyophilized powder, 20-40 units/mg solid