Merck
  • Calculating virtual log P in the alkane/water system (log P(N)(alk)) and its derived parameters deltalog P(N)(oct-alk) and log D(pH)(alk).
Calculating virtual log P in the alkane/water system (log P(N)(alk)) and its derived parameters deltalog P(N)(oct-alk) and log D(pH)(alk).
Journal of medicinal chemistry (2005-04-29)
Giulia Caron, Giuseppe Ermondi
ABSTRACT
Growing interest in the use of both the logarithm of the partition coefficient of the neutral species in the alkane/water system (log P(N)(alk)) and the difference between log P(N)(oct) (the logarithm of the partition coefficient of the neutral species in the n-octanol/water system) and log P(N)(alk) (Deltalog P(N)(oct-alk)) in the early stages of drug design has stimulated development of a computational tool based on the Volsurf software to predict virtual (=of each conformer) log P(N)(alk) and virtual log P(N)(oct). From these two pieces of data, it is then possible to calculate Deltalog P(N)(oct-alk) for a given compound as the difference between log P(N)(oct) and log P(N)(alk). Once the pK(a) is known and the legitimacy of neglecting the contribution made by the ionized species has been checked, it is also possible to calculate log D(pH)(alk), which might be an important lipophilicity descriptor in absorption, distribution, metabolism, and excretion (ADME) prediction, from log P(N)(alk).
Product Description
Supelco
Chloroform solution, certified reference material, 5000 μg/mL in methanol
Supelco
Melting point standard 182-184°C, analytical standard
Sigma-Aldrich
(±)-Sulpiride
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Supelco
2-Hexanone, analytical standard
Sigma-Aldrich
Oxazepam
Sigma-Aldrich
Pindolol, ≥98% (TLC), powder
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Supelco
Warfarin, analytical standard
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Supelco
Testosterone, VETRANAL®, analytical standard
Sigma-Aldrich
Benzaldehyde, puriss. p.a., ≥99.0% (GC)
Supelco
N-Methylaniline, analytical standard
Supelco
Acetone, analytical standard
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Butylamine, ≥99%
Sigma-Aldrich
3-Fluorophenol, 98%
Sigma-Aldrich
Benzaldehyde, ReagentPlus®, ≥99%
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Sigma-Aldrich
p-Anisic acid, ≥99%, FG
Sigma-Aldrich
Benzaldehyde, natural, FCC, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
3-Methoxyphenol, 96%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Benzaldehyde, ≥98%, FG, FCC
Sigma-Aldrich
Acetone, natural, ≥97%