Merck
  • Why gold nanoparticles are more precious than pretty gold: noble metal surface plasmon resonance and its enhancement of the radiative and nonradiative properties of nanocrystals of different shapes.
Why gold nanoparticles are more precious than pretty gold: noble metal surface plasmon resonance and its enhancement of the radiative and nonradiative properties of nanocrystals of different shapes.
Chemical Society reviews (2006-03-01)
Susie Eustis, Mostafa A el-Sayed
ABSTRACT
This tutorial review presents an introduction to the field of noble metal nanoparticles and their current applications. The origin of the surface plasmon resonance and synthesis procedures are described. A number of applications are presented that take advantage of the electromagnetic field enhancement of the radiative properties of noble metal nanoparticles resulting from the surface plasmon oscillations.
Sigma-Aldrich
Gold nanorods, 10 nm diameter, Cy3 and NHS functionalized, powder
Sigma-Aldrich
Gold nanorods, 25 nm diameter, FITC and carboxyl functionalized, powder
Sigma-Aldrich
Gold nanorods, 25 nm diameter, FITC and amine functionalized, powder
Sigma-Aldrich
Gold nanorods, 10 nm diameter, Cy3 and streptavidin functionalized, powder
Sigma-Aldrich
Gold nanorods, 10 nm diameter, Cy3 and amine functionalized, powder
Sigma-Aldrich
Gold nanorods, 10 nm diameter, Cy3 and methyl functionalized, powder
Sigma-Aldrich
Gold nanorods, 25 nm diameter, FITC and NHS functionalized, powder
Sigma-Aldrich
Gold nanorods, 25 nm diameter, FITC and streptavidin functionalized, powder
Sigma-Aldrich
Gold nanorods, 25 nm diameter, FITC and biotin functionalized, powder
Sigma-Aldrich
Gold nanorods, 25 nm diameter, FITC and protein A functionalized, powder
Sigma-Aldrich
Gold nanorods, 10 nm diameter, Cy3 and maleimide functionalized, powder
Sigma-Aldrich
Gold nanorods, 25 nm diameter, FITC and methyl functionalized, powder
Sigma-Aldrich
Gold nanorods, 10 nm diameter, Cy3 and carboxyl functionalized, powder
Sigma-Aldrich
Gold nanorods, 25 nm diameter, FITC and azide functionalized, powder
Sigma-Aldrich
Gold nanorods, 10 nm diameter, Cy3 and protein A functionalized, powder
Sigma-Aldrich
Gold nanorods, 10 nm diameter, Cy3 and azide functionalized, powder
Sigma-Aldrich
Gold nanorods, 10nm diameter, Cy3 and biotin functionalized, powder
Sigma-Aldrich
Gold nanorods, 25 nm diameter, FITC and maleimide functionalized, powder
Sigma-Aldrich
Gold nanospheres, 15 nm diameter, Cy3 and streptavidin functionalized, powder
Sigma-Aldrich
Gold nanospheres, 15 nm, Cy3 and azide functionalized, powder
Sigma-Aldrich
Gold nanospheres, 100 nm, FITC and carboxyl functionalized, powder
Sigma-Aldrich
Gold nanospheres, 100 nm, FITC and maleimide functionalized, powder
Sigma-Aldrich
Gold nanospheres, 100 nm, FITC and methyl functionalized, powder
Sigma-Aldrich
Gold nanospheres, 100 nm, FITC and NHS functionalized, powder
Sigma-Aldrich
Gold nanospheres, 100 nm, FITC and streptavidin functionalized, powder
Sigma-Aldrich
Gold nanospheres, 15 nm, Cy3 and NHS functionalized, powder
Sigma-Aldrich
Gold nanospheres, 100 nm, FITC and biotin functionalized, powder
Sigma-Aldrich
Gold nanospheres, 15 nm, Cy3 and maleimide functionalized, powder
Sigma-Aldrich
Gold nanospheres, 15 nm, Cy3 and methyl functionalized, powder
Sigma-Aldrich
Gold nanospheres, 15 nm, Cy3 and protein A functionalized, powder