Skip to Content
Merck
All Photos(1)

Key Documents

906174

Sigma-Aldrich

Gold nanorods

25 nm diameter, FITC and amine functionalized, powder

Synonym(s):

Dye labeled gold nanoparticles, FITC labeled gold nanorods, FITC tagged gold nanorods, Fluorescent labeled gold nanoparticles, Au nanorods, Gold nanorod

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12141717
NACRES:
NA.23

form

nanorod
powder

diameter

25 nm

storage temp.

2-8°C

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Owing to size and shape dependent quantum effects, the gold nanoparticles exhibit unique surface plasmon resonance absorption, scattering, fluorescence, and photothermal properties which render them suitable for applications in catalysis, chemical sensing, biosensing, cellular and bioimaging, drug and gene delivery and photothermal therapy. The fluorescent labeling of gold nanoparticles have been shown to cause emission enhancement in fluorophores. This makes the fluorescence tagged gold nanoparticles a dual mode nanoprobe for drug delivery and bioimaging applications.

Legal Information

Product of Nanopartz Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorescently Labeled Gold Nanoparticles with Minimal Fluorescence Quenching.
Lu Y, et al.
The Journal of Physical Chemistry C, 114, 17446-17446 (2010)
Byoungjin Kim et al.
Nature nanotechnology, 5(6), 465-472 (2010-04-13)
Nanoparticles have great potential as controllable drug delivery vehicles because of their size and modular functionality. Timing and location are important parameters when optimizing nanoparticles for delivery of chemotherapeutics. Here, we show that gold nanoparticles carrying either fluorescein or doxorubicin
Parwathy Chandran et al.
Nanotoxicology, 11(4), 507-519 (2017-04-20)
This study investigated the role of nanoparticle size and surface chemistry on biocorona composition and its effect on uptake, toxicity and cellular responses in human umbilical vein endothelial cells (HUVEC), employing 40 and 80 nm gold nanoparticles (AuNP) with branched polyethyleneimine
Gold Nanoparticles for In Vitro Diagnostics.
Wen Zhou et al.
Chemical reviews, 115(19), 10575-10636 (2015-06-27)
Xiaohua Huang et al.
Advanced materials (Deerfield Beach, Fla.), 21(48), 4880-4910 (2009-12-28)
Noble metal nanoparticles are capable of confining resonant photons in such a manner as to induce coherent surface plasmon oscillation of their conduction band electrons, a phenomenon leading to two important properties. Firstly, the confinement of the photon to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service