Skip to Content
Merck
All Photos(1)

Key Documents

906204

Sigma-Aldrich

Gold nanorods

25 nm diameter, FITC and methyl functionalized, powder

Synonym(s):

Dye labeled gold nanoparticles, FITC labeled gold nanorods, FITC tagged gold nanorods, Fluorescent labeled gold nanoparticles, Au nanorods, Gold nanorod

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12141717
NACRES:
NA.23

form

nanorod
powder

diameter

25 nm

storage temp.

2-8°C

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Owing to size and shape dependent quantum effects, the gold nanoparticles exhibit unique surface plasmon resonance absorption, scattering, fluorescence, and photothermal properties which render them suitable for applications in catalysis, chemical sensing, biosensing , cellular and bioimaging, drug and gene delivery, and photothermal therapy. The fluorescent labeling of gold nanoparticles have been shown to cause emission enhancement in fluorophore. This makes the fluorescence tagged gold nanoparticles a dual mode nanoprobe for drug delivery and bioimaging applications.

Legal Information

Product of Nanopartz Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorescently Labeled Gold Nanoparticles with Minimal Fluorescence Quenching.
Lu Y, et al.
The Journal of Physical Chemistry C, 114, 17446-17446 (2010)
Gold Nanoparticles for In Vitro Diagnostics.
Wen Zhou et al.
Chemical reviews, 115(19), 10575-10636 (2015-06-27)
Xiaohua Huang et al.
Advanced materials (Deerfield Beach, Fla.), 21(48), 4880-4910 (2009-12-28)
Noble metal nanoparticles are capable of confining resonant photons in such a manner as to induce coherent surface plasmon oscillation of their conduction band electrons, a phenomenon leading to two important properties. Firstly, the confinement of the photon to the
Parwathy Chandran et al.
Nanotoxicology, 11(4), 507-519 (2017-04-20)
This study investigated the role of nanoparticle size and surface chemistry on biocorona composition and its effect on uptake, toxicity and cellular responses in human umbilical vein endothelial cells (HUVEC), employing 40 and 80 nm gold nanoparticles (AuNP) with branched polyethyleneimine
Catherine J Murphy et al.
Accounts of chemical research, 41(12), 1721-1730 (2008-08-21)
Gold, enigmatically represented by the target-like design of its ancient alchemical symbol, has been considered a mystical material of great value for centuries. Nanoscale particles of gold now command a great deal of attention for biomedical applications. Depending on their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service