Merck
  • Determination of lipoic acid in human plasma by HPLC-ECD using liquid-liquid and solid-phase extraction: Method development, validation and optimization of experimental parameters.
Determination of lipoic acid in human plasma by HPLC-ECD using liquid-liquid and solid-phase extraction: Method development, validation and optimization of experimental parameters.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2010-09-11)
Abad Khan, Muhammad I Khan, Zafar Iqbal, Lateef Ahmad, Yasar Shah, David G Watson
ABSTRACT
A rapid, inexpensive, sensitive and specific HPLC-ECD method for the determination of lipoic acid in human plasma was developed and validated over the linearity range of 0.001-10μg/ml using naproxen sodium as an internal standard (IS). Extraction of lipoic acid and IS from plasma (250μl) was carried out with a simple one step liquid-liquid extraction using dichloromethane. Similarly solid-phase extraction was carried out using dichloromethane as extraction solvent. The separated organic layer was dried under the stream of nitrogen at 40°C and the residue was reconstituted with the mobile phase. Complete separation of both lipoic acid and IS at 30°C on Discovery HS C18 RP column (250mm×4.6mm, 5μm) was achieved in 6min using 0.05M phosphate buffer (pH 2.5 adjusted with phosphoric acid):acetonitrile (50:50, v/v) as a mobile phase pumped at the rate of 1.5ml/min using electrochemical detector in DC mode at the detector potential of 1.0V. The limit of detection and limit of quantification of lipoic acid were 200pg/ml and 1ng/ml, respectively. While on column limit of detection and limit of quantification of lipoic acid were 10 and 50pg/ml, respectively. The absolute recoveries of lipoic acid with liquid-liquid and solid-phase extraction were 98.43, 95.65, 101.45, and 97.36, 102.73, 100.17% at 0.5, 1 and 5μg/ml levels, respectively. Coefficient of variations for both intra-day and inter-day were between 0.28 and 4.97%. The method is validated and will be quite suitable for the analysis of lipoic acid in the plasma of human volunteers as well as patients with diabetes and cardiovascular diseases.
Product Description
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 2.1 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 21.2 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Discovery® HS C18 HPLC Column, 10 μm particle size, L × I.D. 25 cm × 21.2 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4 mm
Supelco
Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
Supelco
Discovery® HS C18 Prep Guard Cartridge, 10 μm particle size, L × I.D. 1 cm × 21.2 mm
Supelco
Discovery® HS C18 HPLC Column, 3 μm particle size, L × I.D. 5 cm × 2.1 mm
Supelco
Discovery® HS C18 HPLC Column, 3 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Discovery® HS C18 HPLC Column, 3 μm particle size, L × I.D. 7.5 cm × 2.1 mm
Supelco
Discovery® HS C18 HPLC Column, 3 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Discovery® HS C18 HPLC Column, 3 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Discovery® HS C18 HPLC Column, 3 μm particle size, L × I.D. 7.5 cm × 4.6 mm