Skip to Content
Merck
All Photos(1)

Key Documents

568543-U

Supelco

Discovery® HS C18 (5 µm) HPLC Columns

L × I.D. 25 cm × 21.2 mm HPLC Prep Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

Discovery® HS C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 21.2 mm

material

stainless steel column

Quality Level

Agency

suitable for USP L1

product line

Discovery®

feature

endcapped

manufacturer/tradename

Discovery®

packaging

1 ea of

extent of labeling

20% Carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

25 cm × 21.2 mm

surface area

300 m2/g

surface coverage

3.2 μmol/m2

impurities

<10 ppm metals

matrix

silica gel, high purity, spherical particle platform
fully porous particle

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

120 Å

operating pH range

2-8

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Stable, low bleed for LC-MS applications
  • Scalable from analytical to preparatory
  • Highly stable to ensure excellent run-to-run and lot-to-lot reproducibility
  • Higher hydrophobicity for better resolution of difficult analytes
   

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jolanta Flieger et al.
Journal of separation science, 34(7), 733-739 (2011-03-02)
Highly hydrophilic compounds belonging to biogenic amines were analysed in the reversed-phase system, modified with the addition of ionic liquids: 1-ethyl-3-methyl-imidazolium hexafluorophosphate (EMIM PF(6)) and chaotropic salt NaPF(6) on Discovery HS C18 column at acidic conditions. The effect of the
Swati Jaiswal et al.
Journal of chromatographic science, 55(6), 617-624 (2017-03-24)
Tuberculosis (TB) with human immunodeficiency virus (HIV)/acquired immunodeficiency syndrome represents the most common infectious diseases worldwide. Anti-TB drugs are used concurrently with antiretroviral drug for treatment of TB-HIV co-morbidities. Due to lower risk of interaction with protease inhibitors, rifabutin is
Abad Khan et al.
Talanta, 84(3), 789-801 (2011-04-13)
A new, simple, economical and validated high-performance liquid chromatography linked with electrochemical detector (HPLC-ECD) method has been developed and optimized for different experimental parameters to analyze the most common monothiols and disulfide (cystine, cysteine, homocysteine, methionine, reduced (GSH) and oxidized
Awantika Singh et al.
Rapid communications in mass spectrometry : RCM, 29(6), 485-496 (2015-07-15)
Adhatoda beddomei and Adhatoda vasica are popular Ayurvedic medicinal plants in India, belonging to the family Acanthaceae. Tricyclic quinazoline alkaloids are found to be the most abundant in these plants which are responsible for broad-spectrum medicinal properties. This study aims
Bhavin N Patel et al.
Biomedical chromatography : BMC, 22(11), 1213-1224 (2008-07-25)
A simple, precise and accurate assay for the determination of 6-methoxy-2-naphthylacetic acid (6-MNA), an active metabolite of nabumetone in human plasma, was developed and validated using liquid chromatography-tandem mass spectrometry (LC-MS/MS). The analyte (6-MNA) and propranolol (internal standard, IS) were

Related Content

HPLC DiscoveryBIO HS C18 products. Available in 3µm, 5µm and 10µm particle sizes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service