Skip to Content
Merck

Skip To

D0422

DMEM - high glucose

With sodium bicarbonate, without ʟ-methionine, ʟ-cystine and ʟ-glutamine, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

DME, Dulbecco′s Modified Eagle′s Medium - high glucose, DMEM

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose and sodium bicarbonate, without L-methionine, L-cystine and L-glutamine, liquid, sterile-filtered, suitable for cell culture

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

glucose: high
phenol red: yes
L-glutamine: no
sodium pyruvate: yes

shipped in

ambient

storage temp.

2-8°C

Application

Dulbecco′s Modified Eagle′s Medium (DMEM) is a modification of Basal Medium Eagle (BME) that contains four-fold concentrations of the amino acids and vitamins. The original formulation contained 1000 mg/L of glucose and was used to culture embryonic mouse cells. Since then, it has been modified in several ways to support primary cultures of mouse and chicken cells, as well as a variety of normal and transformed cells. Each of these media offers a different combination of L-glutamine and sodium pyruvate. Additionally, the glucose levels have been raised to 4500 mg/L, contributing to the name "DMEM/High".

Preparation Note

Supplement with 0.584 g/L L-glutamine.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D1145D6171D7777
technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

form

liquid

form

liquid

form

liquid

form

powder

sterility

sterile-filtered

sterility

sterile-filtered

sterility

sterile-filtered

sterility

-

components

glucose: high
phenol red: yes
L-glutamine: no
sodium pyruvate: yes

components

L-glutamine: no
sodium pyruvate: no
glucose: high
phenol red: no
NaHCO3: yes
HEPES: no

components

sodium pyruvate: no
HEPES: 25 mM
glucose: high
NaHCO3: yes
L-glutamine: no
phenol red: yes

components

glucose: high
sodium pyruvate: yes
NaHCO3: no
HEPES: no
phenol red: yes
L-glutamine: yes

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Stacey L Borrego et al.
Journal of lipid research, 62, 100056-100056 (2021-03-02)
Methionine (Met) is an essential amino acid and critical precursor to the cellular methyl donor S-adenosylmethionine. Unlike nontransformed cells, cancer cells have a unique metabolic requirement for Met and are unable to proliferate in growth media where Met is replaced
Ruth I C Glasgow et al.
Neurogenetics, 18(4), 227-235 (2017-10-28)
Mitochondrial diseases are characterised by clinical, molecular and functional heterogeneity, reflecting their bi-genomic control. The nuclear gene GFM2 encodes mtEFG2, a protein with an essential role during the termination stage of mitochondrial translation. We present here two unrelated patients harbouring
Anne Roobol et al.
Metabolic engineering, 59, 98-105 (2020-02-18)
There is a desire to engineer mammalian host cell lines to improve cell growth/biomass accumulation and recombinant biopharmaceutical protein production in industrially relevant cell lines such as the CHOK1 and HEK293 cell lines. The over-expression of individual subunits of the eukaryotic



Global Trade Item Number

SKUGTIN
D0422-100ML04061833557662

Questions

Reviews

No rating value

Active Filters