Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Casein from bovine milk
CAS Number:
9000-71-9
EC Number:
232-555-1
Products
Product Category
proteins (3)
Brand
Sigma-Aldrich (6)
Supelco (1)
Biological Source
Melting Point (°C)
Physical Form
Product Line
Tissue
Quality Segment
Application
Special Grade
Technical (1)
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
C7078
Casein from bovine milk,
powder
Expand
View Pricing
C5890
Casein from bovine milk,
suitable for substrate for protein kinase (after dephosphorylation), purified powder
Expand
View Pricing
C3400
Casein from bovine milk,
powder
Expand
View Pricing
C6554
Casein from bovine milk,
BioReagent, suitable for insect cell culture
Expand
View Pricing
C5679
Casein from bovine milk,
BioReagent, suitable for insect cell culture
Expand
View Pricing
1.02241
Casein from bovine milk,
soluble in alkali, Technipur
®
Expand
View Pricing
C7906
Casein from bovine milk,
for use in the Total Dietary Fiber Control Kit, TDF-C10
Expand
View Pricing