Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
1,5-Diaminonaphthalene
Synonym(s):
1,5-DAN, 1,5-Naphthalenediamine, NSC 401110
Linear Formula:
C
10
H
6
(NH
2
)
2
CAS Number:
2243-62-1
Molecular Weight:
158.20
EC Number:
218-817-8
Products
Product Category
building blocks (1)
Brand
Supelco (1)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Analyte Suitability
Chemical Composition
Quality Segment
Markush Class
Markush Group
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
D21200
1,5-Diaminonaphthalene,
97%
Expand
View Pricing
56451
1,5-Diaminonaphthalene,
matrix substance for MALDI-MS, ≥99.0% (HPLC)
Expand
View Pricing