D21200
1,5-Diaminonaphthalene
97%
Synonym(s):
1,5-DAN, 1,5-Naphthalenediamine, NSC 401110
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
C10H6(NH2)2
CAS Number:
Molecular Weight:
158.20
Beilstein:
907947
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
powder
mp
185-187 °C (lit.)
fluorescence
λex 330 nm (H2O:MeOH (1:4))
SMILES string
Nc1cccc2c(N)cccc12
InChI
1S/C10H10N2/c11-9-5-1-3-7-8(9)4-2-6-10(7)12/h1-6H,11-12H2
InChI key
KQSABULTKYLFEV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
438.8 °F - DIN 51758
Flash Point(C)
226 °C - DIN 51758
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Roger Théberge et al.
Journal of the American Society for Mass Spectrometry, 26(8), 1299-1310 (2015-05-24)
Hemoglobinopathies are the most common inherited disorders in humans and are thus the target of screening programs worldwide. Over the past decade, mass spectrometry (MS) has gained a more important role as a clinical means to diagnose variants, and a
1,5-Naphthalenediamine.
IARC monographs on the evaluation of the carcinogenic risk of chemicals to humans, 27, 127-132 (1982-04-01)
Daniel Malko et al.
Journal of the American Chemical Society, 138(49), 16056-16068 (2016-12-15)
Although major progress has recently been achieved through ex situ methods, there is still a lack of understanding of the behavior of the active center in non-precious metal Fe-N/C catalysts under operating conditions. Utilizing nitrite, nitric oxide, and hydroxylamine as
Miklós Nagy et al.
Molecules (Basel, Switzerland), 25(4) (2020-02-23)
: Multiple drug resistant fungi pose a serious threat to human health, therefore the development of completely new antimycotics is of paramount importance. The in vitro antifungal activity of the original, 1-amino-5-isocyanonaphthalenes (ICANs) was evaluated against reference strains of clinically
Mandana Hunter et al.
Journal of neurochemistry, 146(5), 613-630 (2018-05-29)
The human subventricular zone (SVZ) has a defined cytological and neurochemical architecture, with four constituent laminae that act in concert to support its neurogenic activity. Lipidomic specialisation has previously been demonstrated in the neurologically normal human SVZ, with enrichment of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service