Skip to Content
Merck
All Photos(1)

Key Documents

1601827

USP

Residual Solvent Class 2 - Trichloroethylene

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Trichloroethylene solution, Trichloroethene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

form

solution

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

single component solution

SMILES string

ClC(=CCl)Cl

InChI

1S/C2HCl3/c3-1-2(4)5/h1H

InChI key

XSTXAVWGXDQKEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Residual Solvent Class 2 - Trichloroethylene USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

closed cup - does not flash

Flash Point(C)

closed cup - does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fariha Zaheer et al.
Neurologic clinics, 29(3), 657-665 (2011-08-02)
Multiple genetic and environmental etiologies have been implicated in the pathogenesis of idiopathic Parkinson disease. Recent observations have suggested an association between chronic exposure to trichloroethylene (TCE) and development of clinical parkinsonism. Animal models of TCE exposure have shown nigrostriatal
Hang Zhang et al.
Biochemical and biophysical research communications, 446(2), 590-595 (2014-03-19)
Trichloroethylene (TCE), a major occupational and environmental pollutant, has been recently associated with aberrant epigenetic changes in experimental animals and cultured cells. TCE is known to cause severe hepatotoxicity; however, the association between epigenetic alterations and TCE-induced hepatotoxicity are not
Hideaki Watanabe et al.
Dermatology (Basel, Switzerland), 221(1), 17-22 (2010-04-22)
Patients having a generalised rash with severe liver dysfunction associated with exposure to trichloroethylene (TCE) have been reported mainly in Asian countries. However, no case has been reported in Japan since the 1990s. Here, we describe a case of hypersensitivity
David J Jollow et al.
Critical reviews in toxicology, 39(9), 782-797 (2009-10-27)
Trichloroethylene (TCE) is a widespread environmental contaminant that is carcinogenic when given in high, chronic doses to certain strains of mice and rats. The capacity of TCE to cause cancer in humans is less clear. The current maximum contaminant level
Dominik D Alexander et al.
International archives of occupational and environmental health, 81(2), 127-143 (2007-05-12)
Findings from epidemiologic studies of trichloroethylene (TCE) exposure and liver cancer have been inconsistent. To quantitatively evaluate this association and to examine sources of heterogeneity, we conducted a meta-analysis of occupational studies of TCE exposure and liver/biliary tract cancer. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service