Skip to Content
Merck

1601102

USP

Residual Solvents Mixture - Class 1

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Residual Solvents Mixture - Class 1 USP reference standard intended for use in specified quality tests and assays.

Also used to prepare standard stock solutions during the screening of water-soluble and insoluble articles according to the general chapter of the United States of Pharmacopeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Carc. 1A - Muta. 1B - Ozone 1 - Skin Sens. 1 - STOT RE 1 - STOT RE 2 - STOT RE 2 Inhalation

target_organs

Blood, Liver,Kidney, Nose,Liver,Kidney

wgk

WGK 3

flash_point_f

closed cup

flash_point_c

closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



?467? Residual solvents
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 49(1), 6712-6712 (2022)



Global Trade Item Number

SKUGTIN
1601102-3X1.2ML04065271209220
1601102-3AMP04061838745002