Skip to Content
Merck
All Photos(1)

Key Documents

1267000

USP

Ethylparaben

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Ethyl 4-hydroxybenzoate, Ethylparaben

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CO2C2H5
CAS Number:
Molecular Weight:
166.17
Beilstein:
1101972
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

parabens

manufacturer/tradename

USP

bp

297-298 °C (lit.)

mp

114-117 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCOC(=O)c1ccc(O)cc1

InChI

1S/C9H10O3/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6,10H,2H2,1H3

InChI key

NUVBSKCKDOMJSU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ethylparaben USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Ethotoin Tablets
  • Ethylparaben
  • Ethylparaben Sodium
  • Propylparaben
  • Propylparaben Sodium
  • Quazepam Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frances Orton et al.
Toxicology and applied pharmacology, 278(3), 201-208 (2013-09-24)
Many xenobiotics have been identified as in vitro androgen receptor (AR) antagonists, but information about their ability to produce combined effects at low concentrations is missing. Such data can reveal whether joint effects at the receptor are induced at low
Pankaj Aggarwal et al.
Journal of chromatography. A, 1364, 96-106 (2014-09-07)
Highly cross-linked monolithic networks (i.e., polyethylene glycol diacrylate, PEGDA) synthesized from monomers containing varying ethylene oxide chain lengths were fabricated inside fused silica capillary columns for use in liquid chromatography (LC) of small molecules. Tergitol was used as a surfactant
F Vela-Soria et al.
Journal of chromatography. A, 1371, 39-47 (2014-12-03)
In recent decades, the industrial development has resulted in the appearance of a large amount of new chemicals that are able to produce disorders in the human endocrine system. These substances, so-called endocrine disrupting chemicals (EDCs), include many families of
Szabolcs Fekete et al.
Journal of chromatography. A, 1359, 124-130 (2014-07-30)
In this study, the retention changes induced by frictional heating were evaluated for model small compounds (150-190Da) and a small protein, namely insulin (5.7kDa). For this purpose, the effect of longitudinal temperature gradient caused by frictional heating was experimentally dissociated
Marina Gorga et al.
Journal of chromatography. A, 1352, 29-37 (2014-06-10)
A novel fully automated method based on dual column switching using turbulent flow chromatography followed by liquid chromatography coupled to tandem mass spectrometry (TFC-LC-MS/MS) was applied for the determination of endocrine disruptors (EDCs) and related compounds in sediment and sewage

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service