Skip to Content
Merck
All Photos(1)

Key Documents

PHR1011

Supelco

Ethylparaben

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Ethyl 4-hydroxybenzoate, EP, Ethylparaben

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CO2C2H5
CAS Number:
Molecular Weight:
166.17
Beilstein:
1101972
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
39024522
PubChem Substance ID:
E Number:
E214
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. E2425000
traceable to USP 1267000

API family

parabens

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

297-298 °C (lit.)

mp

114-117 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCOC(=O)c1ccc(O)cc1

InChI

1S/C9H10O3/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6,10H,2H2,1H3

InChI key

NUVBSKCKDOMJSU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Ethylparaben may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by high performance liquid chromatography (HPLC).[1]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3158 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of combined p-hydroxy benzoic acid preservatives in a liquid pharmaceutical formulation by HPLC
Shabir GA.
Journal of Pharmaceutical and Biomedical Analysis, 34(1), 207-213 (2004)
Maw-Rong Lee et al.
Journal of chromatography. A, 1120(1-2), 244-251 (2006-03-04)
This study evaluated supercritical fluid extraction (SFE) combined with liquid chromatography-mass spectrometry (LC-MS) to determine trace preservatives and antioxidants including methylparaben (MP), ethylparaben (EP), propylparaben (PP), butylparaben (BP), butylated hydroxyanisole (BHA), butylated hydroxytoluene (BHT), alpha-tocopherol (alpha-t) and alpha-tocopherol acetate (alpha-ta)
Suzanne Abbas et al.
Drug metabolism and pharmacokinetics, 25(6), 568-577 (2010-10-12)
Parabens (alkyl esters of 4-hydroxybenzoic acid) are widely used as preservatives in drugs, cosmetic products, and foodstuffs. Safety concerns have recently increased due to the potential health risks associated to exposure to large amounts of these substances. Biotransformation of parabens
S Nicoli et al.
Journal of pharmaceutical sciences, 97(11), 4830-4839 (2008-03-05)
This work aims at investigating the nicotinamide (NA)-ethyl-paraben (EP) binary system both in solution and in the solid state. In particular, the apparent EP solubility in water was studied in the presence of different NA concentrations (between 0.28 and 1.64
Sawsan El Hussein et al.
Experimental dermatology, 16(10), 830-836 (2007-09-12)
Concern is continuously raised about the safety of parabens which are present in most of the cosmetic preparations. In this investigation, methyl-, ethyl-, propyl- and butyl paraben (MP, EP, PP, BP), in a commercial cosmetic lotion, were deposited on human

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service