Skip to Content
Merck
All Photos(1)

Documents

807167

TSKgel® CM-2SW (5 µm) HPLC Columns

L x I.D. 25 cm × 4.6 mm, carboxymethyl phase

Synonym(s):

TSKgel® CM-SW Columns

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

product name

TSKgel® CM-2SW HPLC Column, phase carboxymethyl, L × I.D. 25 cm × 4.6 mm, 5 μm particle size

material

stainless steel

product line

TSKgel®

feature

counter ion Na+

manufacturer/tradename

Tosoh CM-2SW, 07167

parameter

≤1 M salt concentration
0-100% organic solvent
1.0 mL/min flow rate
10-45 °C temperature
150 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

25 cm × 4.6 mm

matrix

silica particle platform

matrix active group

carboxymethyl phase

particle size

5 μm

pore size

125 Å

operating pH

2.0-7.5

separation technique

cation exchange

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel CM-2SW columns are packed with 5 micron, high efficiency, spherical silica beads which are surface modified with a weak cation exchanger. They are used for the separation and analysis of small proteins, peptides, and other biologically active molecules. They have a smaller pore size than the TSKgel CM-3SW columns.

Application

TSKgel® CM-2SW HPLC column may be used as a cation exchange column in the purification of ghrelin from goldfish intestine using reversed phase high performance liquid chromatography (RP-HPLC).

Physical form

Shipped in methanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Purification and properties of ghrelin from the intestine of the goldfish, Carassius auratus.
Miura T, et al.
Peptides, 30(4), 758-765 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service