Skip to Content
Merck
All Photos(1)

Documents

59615

Supelco

SUPELCOSIL LC-8 (5 µm) HPLC Columns

L × I.D. 2 cm × 2.1 mm Supelguard Guard Cartridge pkg of 2 ea, Guard Cartridge holder required for use

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

product name

SUPELCOSIL LC-8 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm

Agency

suitable for USP L7

packaging

pkg of 2 ea

technique(s)

HPLC: suitable

L × I.D.

2 cm × 2.1 mm

matrix active group

C8 (octyl) phase

particle size

5 μm

pore size

120 Å

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Borch et al.
Journal of chromatography. A, 1022(1-2), 83-94 (2004-02-03)
Explosives such as 2,4,6-trinitrotoluene (TNT), octahydro-1,3,5,7-tetranitro-1,3,5,7-tetrazocine (HMX), and hexahydro-1,3,5-trinitro-1,3,5-triazine (RDX) are widely distributed environmental contaminants. Complete chromatographic separation is necessary in order to accurately determine and quantify explosives and their degradation products in environmental samples and in (bio)transformation studies. The
Jadwiga Piwowarska et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 805(1), 1-5 (2004-04-29)
A high-performance liquid chromatographic method is described for determination of lidocaine (2-(dietyloamino)-N-(2,6-dimetylofenylo) acetamid) and its metabolite, monoethylglycine xylidide (MEGX), in human serum containing various concentration of bilirubin. Lidocaine and its metabolite were extracted from human serum using dichloromethane. After separation
P K Kunicki
Journal of chromatography. B, Biomedical sciences and applications, 750(1), 41-49 (2001-02-24)
A HPLC-UV determination of clobazam and N-desmethylclobazam in human serum and urine is presented. After simple liquid-liquid extraction with dichloromethane the compounds and an internal standard diazepam were separated on a Supelcosil LC-8-DB column at ambient temperature under isocratic conditions
F Belal et al.
Journal of pharmaceutical and biomedical analysis, 24(3), 335-342 (2001-02-24)
A simple, rapid and sensitive HPLC method has been developed for the simultaneous determination of ramipril and hydrochlorothiazide in their dosage forms. Acetonitrile: sodium perchlorate solution (0.1 M) adjusted to pH 2.5+/-0.2 with phosphoric acid (46:54 v/v), was used as
Vesna Cerkvenik-Flajs et al.
Analytica chimica acta, 663(2), 165-171 (2010-03-09)
An analytical method has been developed for the simultaneous determination of the following endectocide drugs in milk: ivermectin, abamectin, doramectin, moxidectin, eprinomectin, emamectin and nemadectin. Samples were extracted with acetonitrile, purified with solid-phase extraction on a reversed phase C(8), derivatised

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service