Skip to Content
Merck
All Photos(1)

Documents

58991

Supelco

SUPELCOSIL LC-8-DB (3 µm) HPLC Columns

L × I.D. 15 cm × 4.6 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

product name

SUPELCOSIL LC-8-DB HPLC Column, 3 μm particle size, L × I.D. 15 cm × 4.6 mm

Agency

suitable for USP L7

feature

endcapped

manufacturer/tradename

SUPELCOSIL

extent of labeling

6.0% carbon loading

parameter

0-70 °C temperature
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

15 cm × 4.6 mm

surface area

170 m2/g

surface coverage

surface coverage 3.2 μmol/m2

matrix

silica gel, spherical particle platform

matrix active group

C8 (octyl) phase

particle size

3 μm

pore size

120 Å

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

SUPELCOSIL LC-DB phases are specially deactivated for basic compounds. These columns provide less retention, better peak shape, and higher efficiency for organic bases than can be obtained on conventional reversed-phase columns.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jadwiga Piwowarska et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 805(1), 1-5 (2004-04-29)
A high-performance liquid chromatographic method is described for determination of lidocaine (2-(dietyloamino)-N-(2,6-dimetylofenylo) acetamid) and its metabolite, monoethylglycine xylidide (MEGX), in human serum containing various concentration of bilirubin. Lidocaine and its metabolite were extracted from human serum using dichloromethane. After separation
P K Kunicki
Journal of chromatography. B, Biomedical sciences and applications, 750(1), 41-49 (2001-02-24)
A HPLC-UV determination of clobazam and N-desmethylclobazam in human serum and urine is presented. After simple liquid-liquid extraction with dichloromethane the compounds and an internal standard diazepam were separated on a Supelcosil LC-8-DB column at ambient temperature under isocratic conditions
Vesna Cerkvenik-Flajs et al.
Analytica chimica acta, 663(2), 165-171 (2010-03-09)
An analytical method has been developed for the simultaneous determination of the following endectocide drugs in milk: ivermectin, abamectin, doramectin, moxidectin, eprinomectin, emamectin and nemadectin. Samples were extracted with acetonitrile, purified with solid-phase extraction on a reversed phase C(8), derivatised
J H Nichols et al.
Clinical chemistry, 40(7 Pt 1), 1312-1316 (1994-07-01)
This automated assay determines the concentration of the antidepressant fluoxetine (Prozac) and its active metabolite norfluoxetine in serum by reversed-phase HPLC with spectrophotometric detection. Extraction, injection, and quantification are performed by the Gilson Aspec automated sample handler. The patient's specimen
M Molíková et al.
Journal of chromatography. A, 1217(8), 1305-1312 (2010-01-12)
The availability of ionic liquids (ILs) in wide areas of application often results in the requirement on their determination. The attention is also often focused on the knowledge of hydrophobicity as it plays a key role in the biological effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service