Skip to Content
Merck
All Photos(2)

Key Documents

57341-U

Supelco

SPME Fiber Assembly

greener alternative

PDMS, for use with autosampler, Nitinol-core (NIT), 23 ga, 100 μm (PDMS)

Synonym(s):

PDMS SPME Fiber Assembly

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
NACRES:
NB.21

Product Name

SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 23 ga

material

fused silica fiber
plain red hub

needle size

23 ga

packaging

pkg of 3 ×  ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

df

100 μm (PDMS)

technique(s)

solid phase microextraction (SPME): suitable

fiber L

1 cm

matrix active group

PDMS coating

application(s)

food and beverages

compatibility

for analyte group volatiles (MW 80-500)
for use with autosampler

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.

Application

Volatiles

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Kovacevic et al.
Journal of chromatography. A, 918(1), 159-167 (2001-06-14)
The composition of hop essential oil is an important tool for evaluation of hop quality. As each hop variety has a typical essential oil pattern (fingerprint), hop oil analyses can be used to distinguish between hop varieties. The headspace solid-phase
Leandro Wang Hantao et al.
Journal of chromatography. A, 1279, 86-91 (2013-02-12)
In this paper is reported the use of the chromatographic profiles of volatiles to determine disease markers in plants - in this case, leaves of Eucalyptus globulus contaminated by the necrotroph fungus Teratosphaeria nubilosa. The volatile fraction was isolated by
Manuela Cerdán-Calero et al.
Journal of chromatography. A, 1241, 84-95 (2012-04-27)
Neutral volatiles and non-volatile polar compounds (sugars, organics acids and aminoacids) present in Valencia Late orange juice have been analysed by Gas Chromatography coupled to Mass Spectrometry (GC-MS). Before analysis, the neutral volatiles have been extracted by Headspace-Solid Phase Microextraction
A Jiménez et al.
Journal of chromatography. A, 1028(2), 321-324 (2004-03-09)
Solid-phase microextraction was used as a technique for headspace sampling of extra virgin olive oil and virgin olive oil samples with different off-flavours. A 100 microm coated polydimethylsiloxane fiber was used to extract volatile aldehydes, the sampling temperature was 45
David Arroyo et al.
Journal of chromatography. A, 1218(28), 4487-4497 (2011-06-04)
An experimental design optimization is reported of an analytical procedure used in the simultaneous determination of seven non-steroidal anti-inflammatory drugs (NSAIDs) in bovine milk by gas chromatography with mass spectrometry detection (GC-MS). This analytical procedure involves a solid-phase microextraction (SPME)

Articles

The work describes the development of an HS-SPME-GC-MS method to determine 2-chloroethanol as a marker of fumigant ethylene oxide in different kinds of sesame seed samples.

The work describes the development of an HS-SPME-GC-MS method to determine 2-chloroethanol as a marker of fumigant ethylene oxide in different kinds of sesame seed samples.

The work describes the development of an HS-SPME-GC-MS method to determine 2-chloroethanol as a marker of fumigant ethylene oxide in different kinds of sesame seed samples.

The work describes the development of an HS-SPME-GC-MS method to determine 2-chloroethanol as a marker of fumigant ethylene oxide in different kinds of sesame seed samples.

See All

Protocols

SPME offers efficient sample preparation for analyzing terpene/terpenoid profiles in plant essential oils using automation.

SPME offers efficient sample preparation for analyzing terpene/terpenoid profiles in plant essential oils using automation.

SPME offers efficient sample preparation for analyzing terpene/terpenoid profiles in plant essential oils using automation.

SPME offers efficient sample preparation for analyzing terpene/terpenoid profiles in plant essential oils using automation.

Chromatograms

suitable for GC, application for SPME

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service