Skip to Content
Merck
All Photos(2)

Documents

57293-U

Supelco

SPME Fiber Assembly

greener alternative

PDMS/DVB, for use with autosampler, Nitinol-core (NIT), 23 ga, 65 μm (PDMS/DVB)

Synonym(s):

PDMS/DVB SPME Fiber Assembly

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115724
NACRES:
NB.21

product name

SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 23 ga, PDMS/DVB StableFlex, for use with autosampler

material

(Polydimethylsiloxane/Divinylbenzene (PDMS/DVB))
StableFlex fiber
plain pink hub

needle size

23 ga

packaging

pkg of 3 ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

df

65 μm (PDMS/DVB)

technique(s)

solid phase microextraction (SPME): suitable

fiber L

1 cm

matrix active group

PDMS/DVB coating

application(s)

food and beverages

compatibility

for analyte group volatiles, amines, and nitroaromatic compounds (MW 50-300)
for use with autosampler

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.

Application

SPME fiber assembly polydimethylsiloxane/divinylbenzene (PDMS/DVB) has been used for the continuous agitation of the vials comprising the sample after incubation for the analysis of volatiles from pineapple fruits using headspace solid phase microextraction and gas chromatography−mass spectrometry (HS-SPME-GC/MS).

Features and Benefits

Designed for use with Merlin Microseal sealing system; can be used with other septumless systems.

Footnote

Coating bonded to a flexible fused silica core, yielding a more suitable coating on a less breakable fiber. There may be a slight difference in extraction selectivity compared to the same coating on a standard fused silica core. 2cm fiber assembly contails no spring.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influence of harvest maturity and fruit logistics on pineapple (Ananas comosus [L.] Merr.) volatiles assessed by headspace solid phase microextraction and gas chromatography?mass spectrometry (HS-SPME-GC/MS).
Steingass BC, et al.
Food Chemistry, 150, 382-391 (2014)
Jing Cheng et al.
Journal of chromatography. A, 1218(18), 2476-2482 (2011-03-23)
A novel ultrasound-assisted surfactant-enhanced emulsification microextraction (UASEME) technique has been proposed by using low-density extraction solvents. In the proposed technique, Tween 80 and cyclohexane were injected into 5-mL glass test tubes with conical bottoms, containing 5.00 mL of a water
Marisela Pontes et al.
Food chemistry, 134(4), 2509-2520 (2013-02-28)
In this study the effect of the cultivar on the volatile profile of five different banana varieties was evaluated and determined by dynamic headspace solid-phase microextraction (dHS-SPME) combined with one-dimensional gas chromatography-mass spectrometry (1D-GC-qMS). This approach allowed the definition of
C A Groom et al.
Journal of chromatography. A, 909(1), 53-60 (2001-02-24)
The cyclic nitramine explosives hexahydro-1,3,5-trinitro-1,3,5-triazine (RDX) and octahydro-1,3,5,7-tetranitro-1,3,5,7-tetrazine (HMX) were examined in field and microcosm soil samples to determine their patterns of degradation and environmental fates. A number of analytical techniques, including solid-phase microextraction with on-fiber derivatization, gas chromatography-mass spectrometry
Noureddine Yassaa et al.
Journal of chromatography. A, 1141(1), 138-144 (2006-12-19)
In order to develop a valuable method for accurate screening of biogenic emissions from undisturbed living plants or for plant-insect interactions, solid-phase microextraction (SPME) has been combined with dynamic branch enclosure cuvettes and enantioselective GC/MS. The study was conducted at

Articles

A direct immersion SPME method using a new overcoated SPME fiber was developed for the extraction of pesticides from pureed prune baby food.

A direct immersion SPME method using a new overcoated SPME fiber was developed for the extraction of pesticides from pureed prune baby food.

A direct immersion SPME method using a new overcoated SPME fiber was developed for the extraction of pesticides from pureed prune baby food.

A direct immersion SPME method using a new overcoated SPME fiber was developed for the extraction of pesticides from pureed prune baby food.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service